Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSPG4
Entrez Gene ID
1464
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16, NG2
Chromosome Location
Chr.15: 75674322 - 75712848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001897.4 NP_001888.2 9 - 10 5228 110
GenBank mRNA AY359468.1 - 9 - 10 5135 110
X96753.1 - 9 - 10 5135 110

View Full Product Details