The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002491.2 | NP_002482.1 | 1 - 2 | 323 | 137 |
GenBank mRNA | AF035839.1 | - | 1 - 1 | 148 | 182 |
AF047183.1 | - | 1 - 2 | 252 | 138 | |
BC018183.1 | - | 1 - 2 | 271 | 137 | |
BM848639.1 | - | 1 - 2 | 323 | 137 |