The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303503.1 | NP_001290432.1 | 3 - 4 | 342 | 91 |
NM_001303504.1 | NP_001290433.1 | 3 - 4 | 336 | 91 | |
NM_001303506.1 | NP_001290435.1 | 3 - 4 | 395 | 91 | |
NM_002720.2 | NP_002711.1 | 3 - 4 | 336 | 91 | |
XM_006721061.3 | XP_006721124.1 | 3 - 4 | 323 | 91 | |
XM_017023403.1 | XP_016878892.1 | 3 - 4 | 312 | 91 | |
GenBank mRNA | AF097996.1 | - | 3 - 4 | 258 | 91 |
BC001416.2 | - | 3 - 4 | 289 | 91 | |
BG741669.1 | - | 3 - 4 | 367 | 91 | |
BQ220878.1 | - | 3 - 4 | 246 | 91 | |
BT019746.1 | - | 2 - 3 | 154 | 91 | |
X70218.1 | - | 3 - 4 | 289 | 91 |