The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303503.1 | NP_001290432.1 | 4 - 5 | 391 | 85 |
NM_001303506.1 | NP_001290435.1 | 4 - 5 | 444 | 85 | |
NM_001303507.1 | NP_001290436.1 | 3 - 4 | 333 | 85 | |
NM_002720.2 | NP_002711.1 | 4 - 5 | 385 | 85 | |
XM_017023403.1 | XP_016878892.1 | 4 - 5 | 361 | 85 | |
XM_017023404.1 | XP_016878893.1 | 3 - 4 | 297 | 85 | |
GenBank mRNA | AF097996.1 | - | 4 - 5 | 307 | 85 |
AL519181.3 | - | 3 - 4 | 287 | 85 | |
BC001416.2 | - | 4 - 5 | 338 | 85 | |
BG706310.1 | - | 3 - 4 | 336 | 85 | |
BG741669.1 | - | 4 - 5 | 416 | 85 | |
BT019746.1 | - | 3 - 4 | 203 | 85 | |
X70218.1 | - | 4 - 5 | 338 | 85 |