Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A1
Entrez Gene ID
6580
Gene Name
solute carrier family 22 member 1
Gene Aliases
HOCT1, OCT1, oct1_cds
Chromosome Location
Chr.6: 160121789 - 160159201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003057.2 NP_003048.1 4 - 5 950 110
NM_153187.1 NP_694857.1 4 - 5 950 110
XM_005267102.4 XP_005267159.1 4 - 5 992 110
XM_005267103.1 XP_005267160.1 4 - 5 992 110
XM_005267104.4 XP_005267161.1 4 - 5 572 110
XM_005267105.4 XP_005267162.1 4 - 5 524 110
XM_006715552.1 XP_006715615.1 4 - 5 992 110
XM_011536074.2 XP_011534376.1 4 - 5 815 110
GenBank mRNA AK289887.1 - 4 - 5 908 110
BC126364.1 - 4 - 5 916 110
U77086.1 - 4 - 5 898 110
X98332.1 - 4 - 5 917 110

View Full Product Details