Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
Chromosome Location
Chr.2: 227472169 - 227561222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135188.1 NP_001128660.1 6 - 7 1064 146
NM_004504.4 NP_004495.2 6 - 7 1064 146
XM_006712480.3 XP_006712543.1 6 - 7 1067 146
XM_017003954.1 XP_016859443.1 2 - 3 1769 146
GenBank mRNA BC030592.2 - 6 - 7 898 146
BC096272.3 - 6 - 7 864 146
BC096273.3 - 6 - 7 864 146
BC096275.3 - 6 - 7 864 146
KU178123.1 - 6 - 7 814 146
KU178126.1 - 6 - 7 814 146
X89478.1 - 6 - 7 880 146

View Full Product Details