Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFKBIL1
Entrez Gene ID
4795
Gene Name
NFKB inhibitor like 1
Gene Aliases
IKBL, LST1, NFKBIL
Chromosome Location
Chr.6: 31546851 - 31558829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144962.1 NP_001138434.1 3 - 4 619 66
NM_005007.3 NP_004998.3 3 - 4 672 66
GenBank mRNA AF097419.1 - 3 - 4 555 66
AF097420.1 - 3 - 4 555 66
AF097421.1 - 3 - 4 555 66
AF097422.1 - 3 - 4 555 66
AF097423.1 - 3 - 4 555 66
AF097424.1 - 3 - 4 555 66
AF097425.1 - 3 - 4 555 66
AF097426.1 - 3 - 4 555 66
AF097427.1 - 3 - 4 555 66
AF097428.1 - 3 - 4 555 66
AF097429.1 - 3 - 4 555 66
AK291893.1 - 3 - 4 672 66
AK300818.1 - 3 - 4 552 66
AK313888.1 - 3 - 4 607 66
BC105064.1 - 3 - 4 610 66
BC105068.1 - 3 - 4 610 66
BM044744.1 - 3 - 4 654 66
X77909.1 - 3 - 4 621 66

View Full Product Details