The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098504.1 | NP_001091974.1 | 2 - 3 | 538 | 92 |
NM_006386.4 | NP_006377.2 | 2 - 3 | 538 | 92 | |
GenBank mRNA | AB209595.1 | - | 2 - 3 | 494 | 92 |
AK024985.1 | - | 2 - 3 | 506 | 92 | |
AK074224.1 | - | 2 - 3 | 506 | 92 | |
AK303049.1 | - | 2 - 3 | 218 | 92 | |
AL713763.1 | - | 2 - 3 | 492 | 92 | |
AU130891.1 | - | 2 - 3 | 535 | 92 | |
BC000595.2 | - | 2 - 3 | 485 | 92 | |
BP353248.1 | - | 2 - 3 | 538 | 92 | |
CR456432.1 | - | 2 - 3 | 228 | 92 | |
U59321.1 | - | 2 - 3 | 198 | 92 |