Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARID3B
Entrez Gene ID
10620
Gene Name
AT-rich interaction domain 3B
Gene Aliases
BDP, DRIL2
Chromosome Location
Chr.15: 74541177 - 74598131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307939.1 NP_001294868.1 1 - 2 158 58
NM_006465.3 NP_006456.1 1 - 2 158 58
GenBank mRNA AB208830.1 - 1 - 2 77 58
AF116846.1 - 1 - 2 90 58
AK298716.1 - 1 - 2 83 58
AK302034.1 - 1 - 2 158 58
BC041792.1 - 1 - 2 83 58
BC045545.1 - 1 - 2 129 58
BC060824.1 - 1 - 2 71 58
DB455943.1 - 1 - 2 148 58
DC396899.1 - 1 - 2 158 58

View Full Product Details