Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMR3B
Entrez Gene ID
10879
Gene Name
submaxillary gland androgen regulated protein 3B
Gene Aliases
P-B, PBII, PRL3, PROL3, SMR1B
Chromosome Location
Chr.4: 70383078 - 70390244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006685.3 NP_006676.1 2 - 3 199 161
GenBank mRNA BC015327.1 - 2 - 3 134 161
BC094707.1 - 2 - 3 146 161
BX487896.1 - 2 - 3 149 161
CB958582.1 - 2 - 3 202 161
D29833.2 - 2 - 3 136 161

View Full Product Details