The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006849.2 | NP_006840.2 | 8 - 9 | 1296 | 114 |
GenBank mRNA | AK075485.1 | - | 9 - 10 | 1213 | 114 |
AK124931.1 | - | 7 - 8 | 1315 | 114 | |
AK295383.1 | - | 8 - 9 | 1188 | 114 | |
AK308516.1 | - | 7 - 8 | 1244 | 114 | |
BC000537.2 | - | 8 - 9 | 1230 | 114 | |
BC075029.2 | - | 8 - 9 | 1228 | 114 | |
U19948.1 | - | 8 - 9 | 1229 | 114 | |
Z84717.1 | - | 4 - 5 | 1524 | 114 |