The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039476.2 | NP_001034565.1 | 8 - 9 | 1390 | 83 |
NM_001077350.2 | NP_001070818.1 | 11 - 12 | 1288 | 83 | |
NM_001243247.1 | NP_001230176.1 | 12 - 13 | 1425 | 83 | |
NM_001243248.1 | NP_001230177.1 | 10 - 11 | 1488 | 83 | |
NM_001243249.1 | NP_001230178.1 | 9 - 10 | 1520 | 83 | |
GenBank mRNA | AK026990.1 | - | 8 - 9 | 968 | 83 |
AK090717.1 | - | 8 - 9 | 1360 | 83 | |
AK294228.1 | - | 12 - 13 | 1421 | 83 | |
AK298795.1 | - | 9 - 10 | 1082 | 83 | |
AK300736.1 | - | 10 - 11 | 1178 | 83 | |
BC004185.2 | - | 8 - 9 | 1077 | 83 | |
BC012825.2 | - | 8 - 9 | 1077 | 83 | |
BC020392.1 | - | 9 - 10 | 1334 | 83 | |
X90857.1 | - | 11 - 12 | 1449 | 83 |