Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAG3
Entrez Gene ID
10734
Gene Name
stromal antigen 3
Gene Aliases
-
Chromosome Location
Chr.7: 100177563 - 100219334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282716.1 NP_001269645.1 8 - 9 1115 66
NM_001282717.1 NP_001269646.1 8 - 9 1196 66
NM_001282718.1 NP_001269647.1 6 - 7 989 66
NM_012447.3 NP_036579.2 8 - 9 1196 66
XM_011515742.1 XP_011514044.1 8 - 9 1163 66
XM_017011683.1 XP_016867172.1 8 - 9 1043 66
XM_017011684.1 XP_016867173.1 8 - 9 1163 66
XM_017011685.1 XP_016867174.1 8 - 9 1276 66
XM_017011686.1 XP_016867175.1 8 - 9 1043 66
XM_017011687.1 XP_016867176.1 6 - 7 869 66
GenBank mRNA AJ007798.2 - 8 - 9 1116 66
AK292394.1 - 8 - 9 1023 66
AK302587.1 - 5 - 6 607 66
AK302693.1 - 6 - 7 812 66
AL833816.1 - 7 - 8 1287 66
BC140932.1 - 8 - 9 955 66
BC146806.1 - 8 - 9 1021 66

View Full Product Details