The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012473.3 | NP_036605.2 | 1 - 2 | 84 | 59 |
XM_005261508.1 | XP_005261565.1 | 1 - 2 | 469 | 59 | |
XM_011530111.2 | XP_011528413.1 | 1 - 2 | 469 | 59 | |
GenBank mRNA | AF480262.1 | - | 1 - 2 | 84 | 59 |
AK301837.1 | - | 1 - 2 | 475 | 59 | |
BC013726.1 | - | 1 - 2 | 45 | 59 | |
BC050610.1 | - | 1 - 2 | 58 | 59 | |
CR456601.2 | - | 1 - 2 | 122 | 59 |