The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019102.3 | NP_061975.2 | 1 - 2 | 627 | 127 |
GenBank mRNA | BC013682.1 | - | 1 - 2 | 614 | 127 |
BI822268.1 | - | 1 - 2 | 126 | ||
BT009744.1 | - | 1 - 2 | 566 | 127 | |
CR542256.1 | - | 1 - 2 | 566 | 127 |