Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM10
Entrez Gene ID
56980
Gene Name
PR domain 10
Gene Aliases
PFM7
Chromosome Location
Chr.11: 129899706 - 130003097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020228.2 NP_064613.2 9 - 10 1390 97
NM_199437.1 NP_955469.1 9 - 10 1390 97
NM_199438.1 NP_955470.1 5 - 6 1116 97
NM_199439.1 NP_955471.1 5 - 6 1116 97
XM_011542912.2 XP_011541214.1 9 - 10 1652 97
XM_017018023.1 XP_016873512.1 5 - 6 3201 97
GenBank mRNA AB033057.2 - 5 - 6 900 97
AF275817.1 - 5 - 6 1116 97
AF503171.1 - 5 - 6 1047 97
AK310354.1 - 10 - 11 1436 97
AL833172.1 - 9 - 10 1393 97
BC112934.1 - 9 - 10 1382 97
BC117415.1 - 5 - 6 955 97
BC143612.1 - 5 - 6 955 97
BC143613.1 - 5 - 6 955 97
BX648944.1 - 9 - 10 1391 97

View Full Product Details