The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317734.1 | NP_001304663.1 | 8 - 9 | 911 | 72 |
NM_001317735.1 | NP_001304664.1 | 6 - 7 | 580 | 72 | |
NM_020524.3 | NP_065385.2 | 9 - 10 | 1000 | 72 | |
GenBank mRNA | AF221521.1 | - | 9 - 10 | 949 | 72 |
AK022497.1 | - | 8 - 9 | 870 | 72 | |
AK023219.1 | - | 9 - 10 | 941 | 72 | |
AK300987.1 | - | 5 - 6 | 289 | 72 | |
AK303413.1 | - | 6 - 7 | 524 | 72 | |
AK303863.1 | - | 6 - 7 | 425 | 72 | |
AK315551.1 | - | 9 - 10 | 981 | 72 | |
BC016852.1 | - | 9 - 10 | 928 | 72 |