Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM8A
Entrez Gene ID
58986
Gene Name
transmembrane protein 8A
Gene Aliases
M83, TMEM6, TMEM8
Chromosome Location
Chr.16: 370776 - 381950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021259.2 NP_067082.2 7 - 8 1444 67
GenBank mRNA AB045292.1 - 7 - 8 1401 67
AK024725.1 - 4 - 5 743 67
AK091823.1 - 7 - 8 1222 67
AL546643.3 - 3 - 4 499 67
BC021557.1 - 7 - 8 1335 67

View Full Product Details