The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022450.3 | NP_071895.3 | 2 - 3 | 266 | 72 |
XM_005255494.1 | XP_005255551.1 | 2 - 3 | 245 | 72 | |
XM_005255498.2 | XP_005255555.1 | 1 - 2 | 120 | 72 | |
GenBank mRNA | AK026010.1 | - | 2 - 3 | 230 | 72 |
AK225408.1 | - | 2 - 3 | 230 | 72 | |
AK291177.1 | - | 2 - 3 | 266 | 72 | |
AK304797.1 | - | 2 - 3 | 242 | 72 | |
AK309097.1 | - | 2 - 3 | 193 | 72 | |
BC014425.1 | - | 2 - 3 | 228 | 72 |