Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMND5B
Entrez Gene ID
64777
Gene Name
required for meiotic nuclear division 5 homolog B
Gene Aliases
GID2, GID2B
Chromosome Location
Chr.5: 178130532 - 178148570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288794.1 NP_001275723.1 7 - 8 919 84
NM_001288795.1 NP_001275724.1 6 - 7 768 84
NM_022762.4 NP_073599.2 6 - 7 779 84
XM_005265969.3 XP_005266026.1 5 - 6 599 84
XM_005265971.4 XP_005266028.1 6 - 7 1346 84
XM_005265972.3 XP_005266029.1 5 - 6 726 84
XM_017009727.1 XP_016865216.1 5 - 6 1390 84
XM_017009728.1 XP_016865217.1 5 - 6 1057 84
GenBank mRNA AK025971.1 - 3 - 4 324 84
AK093481.1 - 6 - 7 720 84
AK125337.1 - 7 - 8 866 84
AL713670.1 - 5 - 6 642 84
AY359092.1 - 3 - 4 1637 84
BC009911.2 - 6 - 7 713 84
DQ494789.1 - 6 - 7 713 84

View Full Product Details