The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024637.4 | NP_078913.3 | 3 - 4 | 828 | 87 |
GenBank mRNA | AF289575.1 | - | 2 - 3 | 623 | 87 |
AF316113.1 | - | 3 - 4 | 671 | 87 | |
AK092367.1 | - | 2 - 3 | 1454 | 87 | |
AK225890.1 | - | 2 - 3 | 604 | 87 | |
AL833824.1 | - | 2 - 3 | 473 | 87 | |
BC012976.2 | - | 3 - 4 | 647 | 87 |