The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052889.2 | NP_443121.1 | 3 - 4 | 359 | 116 |
GenBank mRNA | AF367017.1 | - | 2 - 3 | 332 | 117 |
BC070196.1 | - | 3 - 4 | 359 | 116 | |
BC104635.1 | - | 3 - 4 | 363 | 116 |