The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139076.2 | NP_620775.2 | 8 - 9 | 856 | 96 |
XM_005263280.4 | XP_005263337.1 | 9 - 10 | 989 | 96 | |
XM_017008694.1 | XP_016864183.1 | 7 - 8 | 762 | 96 | |
GenBank mRNA | AK021582.1 | - | 3 - 4 | 1064 | 96 |
AK022704.1 | - | 8 - 9 | 842 | 96 | |
AK023676.1 | - | 7 - 8 | 756 | 96 | |
AY358576.1 | - | 8 - 9 | 807 | 96 | |
BC039573.1 | - | 8 - 9 | 803 | 96 | |
EF531340.1 | - | 8 - 9 | 791 | 96 |