The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145905.2 | NP_665912.1 | 1 - 2 | 169 | 101 |
GenBank mRNA | BC015789.1 | - | 1 - 2 | 169 | 101 |