Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58703140-58703595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51C
Entrez Gene ID
5889
Gene Name
RAD51 paralog C
Gene Aliases
BROVCA3, FANCO, R51H3, RAD51L2
UniGene
Hs.412587
Gene Chromosome Location
Chr.17: 58692538-58734342 on Build GRCh38
Transcripts
NM_058216.2 NR_103872.1 XM_006722001.3 XM_006722002.3 XM_006722004.3 XM_006722005.3 XM_011525092.2 XM_011525093.2 XM_011525094.2 XM_017024914.1 XM_017024915.1 XM_017024916.1 XM_017024917.1 XM_017024918.1 XM_017024919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303122 c.670T>C p.Y224H Chr.17: 58703294-58703294 on Build GRCh38
6982831 c.703A>C p.K235Q Chr.17: 58703327-58703327 on Build GRCh38
6917715 c.626A>G p.Y209C Chr.17: 58703250-58703250 on Build GRCh38
6912127 c.605A>T p.D202V Chr.17: 58703229-58703229 on Build GRCh38
7095185 c.687C>T p.F229F Chr.17: 58703311-58703311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770096766 Chr.17: 58703203-58703203 on Build GRCh38
rs746890699 Chr.17: 58703185-58703185 on Build GRCh38
rs899177307 Chr.17: 58703408-58703408 on Build GRCh38
rs949601972 Chr.17: 58703360-58703360 on Build GRCh38
rs587781921 Chr.17: 58703244-58703244 on Build GRCh38
rs746755735 Chr.17: 58703271-58703271 on Build GRCh38
rs758958193 Chr.17: 58703361-58703362 on Build GRCh38
rs982591070 Chr.17: 58703366-58703366 on Build GRCh38
rs193023469 Chr.17: 58703179-58703179 on Build GRCh38
rs200857129 Chr.17: 58703259-58703259 on Build GRCh38
rs771078849 Chr.17: 58703246-58703246 on Build GRCh38
rs950606402 Chr.17: 58703484-58703484 on Build GRCh38
rs137947462 Chr.17: 58703258-58703258 on Build GRCh38
rs1032681872 Chr.17: 58703298-58703298 on Build GRCh38
rs573610430 Chr.17: 58703498-58703498 on Build GRCh38
rs587780258 Chr.17: 58703185-58703185 on Build GRCh38
rs999954107 Chr.17: 58703562-58703562 on Build GRCh38
rs730881924 Chr.17: 58703295-58703295 on Build GRCh38
rs786201177 Chr.17: 58703320-58703320 on Build GRCh38
rs786203081 Chr.17: 58703277-58703278 on Build GRCh38
rs780177888 Chr.17: 58703311-58703311 on Build GRCh38
rs757896995 Chr.17: 58703181-58703181 on Build GRCh38
rs587780839 Chr.17: 58703226-58703226 on Build GRCh38
rs538884532 Chr.17: 58703231-58703231 on Build GRCh38
rs750060665 Chr.17: 58703272-58703272 on Build GRCh38
rs145310733 Chr.17: 58703194-58703194 on Build GRCh38
rs200293302 Chr.17: 58703201-58703201 on Build GRCh38
rs755849719 Chr.17: 58703329-58703329 on Build GRCh38
rs185219570 Chr.17: 58703382-58703382 on Build GRCh38
rs1008767172 Chr.17: 58703500-58703500 on Build GRCh38
rs748043225 Chr.17: 58703328-58703328 on Build GRCh38
rs900214818 Chr.17: 58703541-58703541 on Build GRCh38
rs786201848 Chr.17: 58703245-58703245 on Build GRCh38
rs775852901 Chr.17: 58703368-58703368 on Build GRCh38
rs377586976 Chr.17: 58703339-58703339 on Build GRCh38
rs774528163 Chr.17: 58703351-58703351 on Build GRCh38
rs745917131 Chr.17: 58703352-58703352 on Build GRCh38
rs775006411 Chr.17: 58703248-58703248 on Build GRCh38
rs964351755 Chr.17: 58703557-58703557 on Build GRCh38
rs544306649 Chr.17: 58703513-58703513 on Build GRCh38
rs876659497 Chr.17: 58703246-58703247 on Build GRCh38
rs757915374 Chr.17: 58703275-58703275 on Build GRCh38
rs878855179 Chr.17: 58703332-58703332 on Build GRCh38
rs773035374 Chr.17: 58703174-58703179 on Build GRCh38
rs1017384420 Chr.17: 58703250-58703250 on Build GRCh38
rs587782332 Chr.17: 58703202-58703202 on Build GRCh38
rs786201909 Chr.17: 58703254-58703254 on Build GRCh38
rs201529791 Chr.17: 58703280-58703280 on Build GRCh38
rs896393923 Chr.17: 58703366-58703367 on Build GRCh38
rs778157321 Chr.17: 58703333-58703333 on Build GRCh38
rs772670037 Chr.17: 58703358-58703358 on Build GRCh38
rs754895072 Chr.17: 58703322-58703322 on Build GRCh38
rs760911964 Chr.17: 58703265-58703265 on Build GRCh38
rs771168730 Chr.17: 58703343-58703343 on Build GRCh38
rs140804406 Chr.17: 58703264-58703264 on Build GRCh38
rs933464572 Chr.17: 58703176-58703176 on Build GRCh38
rs553830720 Chr.17: 58703464-58703464 on Build GRCh38
rs372386600 Chr.17: 58703402-58703402 on Build GRCh38
rs751528808 Chr.17: 58703312-58703312 on Build GRCh38
rs561703592 Chr.17: 58703389-58703389 on Build GRCh38
rs780022466 Chr.17: 58703184-58703184 on Build GRCh38
rs531838785 Chr.17: 58703225-58703225 on Build GRCh38
rs751241015 Chr.17: 58703274-58703275 on Build GRCh38
rs958242201 Chr.17: 58703303-58703303 on Build GRCh38
rs1047665143 Chr.17: 58703455-58703455 on Build GRCh38
rs876658644 Chr.17: 58703330-58703330 on Build GRCh38
rs876660177 Chr.17: 58703291-58703291 on Build GRCh38
rs929193795 Chr.17: 58703414-58703414 on Build GRCh38
rs778403943 Chr.17: 58703177-58703177 on Build GRCh38
rs926760865 Chr.17: 58703375-58703375 on Build GRCh38
rs587782818 Chr.17: 58703325-58703325 on Build GRCh38
rs587782286 Chr.17: 58703275-58703276 on Build GRCh38
rs756889526 Chr.17: 58703174-58703174 on Build GRCh38

View Full Product Details