Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132084231-132084789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs891149268 Chr.X: 132084476-132084476 on Build GRCh38
rs924854606 Chr.X: 132084394-132084394 on Build GRCh38
rs990301741 Chr.X: 132084427-132084427 on Build GRCh38
rs915486972 Chr.X: 132084591-132084591 on Build GRCh38
rs778132971 Chr.X: 132084652-132084652 on Build GRCh38
rs137852212 Chr.X: 132084546-132084546 on Build GRCh38
rs754878275 Chr.X: 132084467-132084467 on Build GRCh38
rs1026735278 Chr.X: 132084412-132084412 on Build GRCh38
rs760380992 Chr.X: 132084523-132084523 on Build GRCh38
rs753587760 Chr.X: 132084461-132084461 on Build GRCh38
rs192276683 Chr.X: 132084307-132084307 on Build GRCh38
rs184868527 Chr.X: 132084758-132084758 on Build GRCh38
rs747204972 Chr.X: 132084447-132084447 on Build GRCh38
rs771052446 Chr.X: 132084450-132084450 on Build GRCh38
rs944037077 Chr.X: 132084639-132084639 on Build GRCh38
rs776019246 Chr.X: 132084490-132084490 on Build GRCh38
rs765724332 Chr.X: 132084624-132084624 on Build GRCh38
rs387906720 Chr.X: 132084540-132084540 on Build GRCh38
rs766009071 Chr.X: 132084527-132084527 on Build GRCh38
rs945648502 Chr.X: 132084665-132084665 on Build GRCh38
rs868145837 Chr.X: 132084499-132084499 on Build GRCh38
rs564880982 Chr.X: 132084741-132084741 on Build GRCh38
rs1016788431 Chr.X: 132084321-132084321 on Build GRCh38
rs773037705 Chr.X: 132084513-132084513 on Build GRCh38
rs961306588 Chr.X: 132084388-132084388 on Build GRCh38
rs761568341 Chr.X: 132084493-132084493 on Build GRCh38
rs760102414 Chr.X: 132084571-132084571 on Build GRCh38
rs934859635 Chr.X: 132084419-132084419 on Build GRCh38
rs749276170 Chr.X: 132084457-132084457 on Build GRCh38
rs747271996 Chr.X: 132084683-132084683 on Build GRCh38
rs34028487 Chr.X: 132084724-132084725 on Build GRCh38
rs761203638 Chr.X: 132084645-132084645 on Build GRCh38
rs753244421 Chr.X: 132084631-132084631 on Build GRCh38
rs991583356 Chr.X: 132084261-132084261 on Build GRCh38
rs1040299822 Chr.X: 132084657-132084657 on Build GRCh38
rs972031615 Chr.X: 132084396-132084396 on Build GRCh38
rs993738599 Chr.X: 132084762-132084762 on Build GRCh38
rs771987980 Chr.X: 132084507-132084507 on Build GRCh38
rs951792457 Chr.X: 132084251-132084251 on Build GRCh38
rs781203168 Chr.X: 132084451-132084451 on Build GRCh38
rs753221288 Chr.X: 132084562-132084562 on Build GRCh38
rs778037459 Chr.X: 132084444-132084444 on Build GRCh38

View Full Product Details