Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CLN8 | NM_018941.3 | NP_061764.2 | ||
XM_005266021.3 | XP_005266078.1 | |||
XM_005266022.1 | XP_005266079.1 | |||
XM_005266023.1 | XP_005266080.1 | |||
XM_011534745.1 | XP_011533047.1 | |||
XM_011534746.2 | XP_011533048.1 | |||
AF123757.1 | AAF13115.1 | |||
AF123758.1 | AAF13116.1 | |||
AF123759.1 | AAF13117.1 | |||
AF123760.1 | AAF13118.1 | |||
AF123761.1 | AAF13119.1 | |||
AK312486.1 | ||||
BC007725.2 | AAH07725.1 | |||
BT007049.1 | AAP35698.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1024325 | Chr8:651043 - 3203837 on Build GRCh38 | Gain | LOC101927752 KBTBD11 MYOM2 LOC101928058 LOC401442 ERICH1-AS1 ARHGEF10 LOC105377774 ERICH1 LOC286083 LOC100131395 DLGAP2-AS1 LOC105377792 KBTBD11-OT1 MIR7160 MIR3674 CSMD1 CLN8 LOC105377781 MIR596 LOC105377776 LOC105377777 LOC101927815 DLGAP2 |
esv3891267 | Chr8:1755403 - 2343278 on Build GRCh38 | Gain | LOC101927752 KBTBD11 MYOM2 KBTBD11-OT1 MIR7160 LOC101928058 MIR3674 CLN8 LOC105377781 ARHGEF10 MIR596 LOC100131395 |
esv3615841 | Chr8:1705527 - 1822229 on Build GRCh38 | Loss | LOC101927752 MIR3674 CLN8 MIR596 DLGAP2 |
dgv11822n54 | Chr8:1748920 - 1829405 on Build GRCh38 | Loss | LOC101927752 MIR3674 CLN8 ARHGEF10 MIR596 |
nsv609527 | Chr8:1733567 - 1805331 on Build GRCh38 | Loss | LOC101927752 MIR3674 CLN8 |
esv3576504 | Chr8:951237 - 1906751 on Build GRCh38 | Gain | LOC101927752 MIR3674 ERICH1-AS1 CLN8 ARHGEF10 MIR596 LOC286083 LOC105377776 DLGAP2-AS1 DLGAP2 |
nsv609531 | Chr8:1768870 - 2267169 on Build GRCh38 | Gain | KBTBD11 MYOM2 KBTBD11-OT1 MIR7160 LOC101928058 MIR3674 CLN8 LOC105377781 ARHGEF10 MIR596 LOC100131395 |
nsv509244 | Chr8:1756513 - 1850704 on Build GRCh38 | Insertion | LOC101927752 MIR3674 CLN8 ARHGEF10 MIR596 |
nsv1025591 | Chr8:1755786 - 2335806 on Build GRCh38 | Gain | LOC101927752 KBTBD11 MYOM2 KBTBD11-OT1 MIR7160 LOC101928058 MIR3674 CLN8 LOC105377781 ARHGEF10 MIR596 LOC100131395 |
esv3615846 | Chr8:1735209 - 1816717 on Build GRCh38 | Loss | LOC101927752 MIR3674 CLN8 |