Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27367702-27368268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7144530 c.778A>G p.M260V Chr.2: 27367750-27367750 on Build GRCh38
3682627 c.651C>G p.S217R Chr.2: 27368139-27368139 on Build GRCh38
7156709 c.708G>A p.V236V Chr.2: 27367820-27367820 on Build GRCh38
4952771 c.788C>G p.P263R Chr.2: 27367800-27367800 on Build GRCh38
5505724 c.653T>G p.L218R Chr.2: 27368077-27368077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763920629 Chr.2: 27367869-27367869 on Build GRCh38
rs35132016 Chr.2: 27367881-27367882 on Build GRCh38
rs990557179 Chr.2: 27367975-27367975 on Build GRCh38
rs931924982 Chr.2: 27367801-27367801 on Build GRCh38
rs770476312 Chr.2: 27367825-27367825 on Build GRCh38
rs992556907 Chr.2: 27367954-27367954 on Build GRCh38
rs968944848 Chr.2: 27367911-27367911 on Build GRCh38
rs372192935 Chr.2: 27367731-27367731 on Build GRCh38
rs775143008 Chr.2: 27367914-27367914 on Build GRCh38
rs558941804 Chr.2: 27367929-27367929 on Build GRCh38
rs940528953 Chr.2: 27367771-27367771 on Build GRCh38
rs770574434 Chr.2: 27368082-27368082 on Build GRCh38
rs1050304821 Chr.2: 27368208-27368208 on Build GRCh38
rs760641987 Chr.2: 27368102-27368102 on Build GRCh38
rs767457541 Chr.2: 27367830-27367830 on Build GRCh38
rs751780070 Chr.2: 27368152-27368152 on Build GRCh38
rs754887517 Chr.2: 27367722-27367722 on Build GRCh38
rs769277328 Chr.2: 27367787-27367787 on Build GRCh38
rs750664457 Chr.2: 27367977-27367977 on Build GRCh38
rs113994027 Chr.2: 27368047-27368047 on Build GRCh38
rs113994029 Chr.2: 27368105-27368105 on Build GRCh38
rs766717475 Chr.2: 27368136-27368136 on Build GRCh38
rs972532369 Chr.2: 27367968-27367968 on Build GRCh38
rs752440296 Chr.2: 27367750-27367750 on Build GRCh38
rs764109646 Chr.2: 27367854-27367854 on Build GRCh38
rs757694836 Chr.2: 27368056-27368056 on Build GRCh38
rs199682969 Chr.2: 27368188-27368188 on Build GRCh38
rs753903529 Chr.2: 27368001-27368002 on Build GRCh38
rs371753289 Chr.2: 27367857-27367857 on Build GRCh38
rs748998115 Chr.2: 27368076-27368076 on Build GRCh38
rs775331791 Chr.2: 27368100-27368100 on Build GRCh38
rs781570273 Chr.2: 27368046-27368046 on Build GRCh38
rs758593908 Chr.2: 27367770-27367770 on Build GRCh38
rs113271372 Chr.2: 27367938-27367938 on Build GRCh38
rs554242828 Chr.2: 27367890-27367890 on Build GRCh38
rs751690179 Chr.2: 27367989-27367989 on Build GRCh38
rs1053260361 Chr.2: 27368240-27368240 on Build GRCh38
rs1028759633 Chr.2: 27367881-27367881 on Build GRCh38
rs747580846 Chr.2: 27367786-27367786 on Build GRCh38
rs186886418 Chr.2: 27367907-27367907 on Build GRCh38
rs750621545 Chr.2: 27367761-27367761 on Build GRCh38
rs537332201 Chr.2: 27367917-27367917 on Build GRCh38
rs767601647 Chr.2: 27368167-27368167 on Build GRCh38
rs753667257 Chr.2: 27368130-27368130 on Build GRCh38
rs756631265 Chr.2: 27368181-27368181 on Build GRCh38
rs949043662 Chr.2: 27368021-27368021 on Build GRCh38
rs755137090 Chr.2: 27368156-27368156 on Build GRCh38
rs1019449767 Chr.2: 27367920-27367920 on Build GRCh38
rs753194689 Chr.2: 27368179-27368179 on Build GRCh38
rs200307171 Chr.2: 27368138-27368138 on Build GRCh38
rs907630897 Chr.2: 27367974-27367974 on Build GRCh38
rs758683857 Chr.2: 27367980-27367980 on Build GRCh38
rs560111940 Chr.2: 27368091-27368091 on Build GRCh38
rs544654144 Chr.2: 27367899-27367899 on Build GRCh38
rs71441072 Chr.2: 27368242-27368242 on Build GRCh38
rs113994030 Chr.2: 27367800-27367800 on Build GRCh38
rs752633032 Chr.2: 27367854-27367854 on Build GRCh38
rs770144131 Chr.2: 27367802-27367802 on Build GRCh38
rs757260632 Chr.2: 27368183-27368184 on Build GRCh38
rs113994028 Chr.2: 27368104-27368104 on Build GRCh38
rs980385158 Chr.2: 27367912-27367912 on Build GRCh38
rs780262752 Chr.2: 27367772-27367772 on Build GRCh38
rs373166860 Chr.2: 27367876-27367876 on Build GRCh38
rs141722828 Chr.2: 27367756-27367756 on Build GRCh38
rs1045950009 Chr.2: 27367829-27367829 on Build GRCh38
rs916241714 Chr.2: 27367979-27367979 on Build GRCh38
rs755881182 Chr.2: 27367751-27367751 on Build GRCh38
rs376346942 Chr.2: 27368073-27368073 on Build GRCh38
rs1034013146 Chr.2: 27367916-27367916 on Build GRCh38
rs944048086 Chr.2: 27368235-27368235 on Build GRCh38
rs931013751 Chr.2: 27367727-27367727 on Build GRCh38
rs907448099 Chr.2: 27368057-27368057 on Build GRCh38
rs780352685 Chr.2: 27367988-27367988 on Build GRCh38
rs577067377 Chr.2: 27367969-27367969 on Build GRCh38
rs773935313 Chr.2: 27368089-27368089 on Build GRCh38
rs778177199 Chr.2: 27368075-27368075 on Build GRCh38
rs911189093 Chr.2: 27368227-27368227 on Build GRCh38
rs949001155 Chr.2: 27367807-27367807 on Build GRCh38
rs41288829 Chr.2: 27367799-27367799 on Build GRCh38
rs748646310 Chr.2: 27368048-27368048 on Build GRCh38
rs1010838120 Chr.2: 27367886-27367886 on Build GRCh38
rs1031796958 Chr.2: 27367873-27367873 on Build GRCh38
rs536216701 Chr.2: 27367877-27367877 on Build GRCh38
rs768451187 Chr.2: 27368175-27368175 on Build GRCh38
rs755047066 Chr.2: 27368030-27368030 on Build GRCh38
rs773837208 Chr.2: 27367826-27367826 on Build GRCh38
rs1049706409 Chr.2: 27367803-27367803 on Build GRCh38
rs151290164 Chr.2: 27367804-27367804 on Build GRCh38
rs572469370 Chr.2: 27368219-27368219 on Build GRCh38

View Full Product Details