Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 88809495-88810094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APRT
Entrez Gene ID
353
Gene Name
adenine phosphoribosyltransferase
Gene Aliases
AMP, APRTD
UniGene
Hs.28914
Gene Chromosome Location
Chr.16: 88809469-88811934 on Build GRCh38
Transcripts
NM_000485.2 NM_001030018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4784557 c.492C>T p.G164G Chr.16: 88809749-88809749 on Build GRCh38
974779 c.388C>T p.L130L Chr.16: 88810082-88810082 on Build GRCh38
250838 c.533A>G p.Q178R Chr.16: 88809708-88809708 on Build GRCh38
974780 c.388C>T p.L130L Chr.16: 88810082-88810082 on Build GRCh38
1709506 c.515C>T p.P172L Chr.16: 88809726-88809726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765012459 Chr.16: 88809658-88809659 on Build GRCh38
rs768425517 Chr.16: 88809750-88809750 on Build GRCh38
rs373775974 Chr.16: 88809740-88809740 on Build GRCh38
rs755380873 Chr.16: 88809711-88809715 on Build GRCh38
rs944384557 Chr.16: 88809622-88809622 on Build GRCh38
rs771629340 Chr.16: 88809899-88809899 on Build GRCh38
rs781239221 Chr.16: 88809727-88809727 on Build GRCh38
rs766563862 Chr.16: 88809941-88809941 on Build GRCh38
rs62048258 Chr.16: 88810025-88810025 on Build GRCh38
rs369507525 Chr.16: 88809788-88809788 on Build GRCh38
rs769047691 Chr.16: 88810044-88810044 on Build GRCh38
rs1056892598 Chr.16: 88809886-88809888 on Build GRCh38
rs751875229 Chr.16: 88809861-88809861 on Build GRCh38
rs137965502 Chr.16: 88809546-88809546 on Build GRCh38
rs2070256 Chr.16: 88809695-88809695 on Build GRCh38
rs776013459 Chr.16: 88809913-88809913 on Build GRCh38
rs772207278 Chr.16: 88809582-88809582 on Build GRCh38
rs147688262 Chr.16: 88809807-88809807 on Build GRCh38
rs914366429 Chr.16: 88809996-88809996 on Build GRCh38
rs752347890 Chr.16: 88809541-88809541 on Build GRCh38
rs547319605 Chr.16: 88809707-88809707 on Build GRCh38
rs777442847 Chr.16: 88809901-88809901 on Build GRCh38
rs752346720 Chr.16: 88809920-88809920 on Build GRCh38
rs752975128 Chr.16: 88809697-88809697 on Build GRCh38
rs8191497 Chr.16: 88809651-88809651 on Build GRCh38
rs769693316 Chr.16: 88809730-88809730 on Build GRCh38
rs372494973 Chr.16: 88809884-88809884 on Build GRCh38
rs387906584 Chr.16: 88809699-88809699 on Build GRCh38
rs1004971357 Chr.16: 88809594-88809594 on Build GRCh38
rs989930459 Chr.16: 88809853-88809853 on Build GRCh38
rs760879511 Chr.16: 88809939-88809939 on Build GRCh38
rs756939078 Chr.16: 88809562-88809562 on Build GRCh38
rs745872435 Chr.16: 88809802-88809802 on Build GRCh38
rs370273203 Chr.16: 88809959-88809959 on Build GRCh38
rs565636938 Chr.16: 88809712-88809712 on Build GRCh38
rs763442250 Chr.16: 88809915-88809915 on Build GRCh38
rs763752678 Chr.16: 88809675-88809675 on Build GRCh38
rs761192074 Chr.16: 88809683-88809683 on Build GRCh38
rs756627246 Chr.16: 88809554-88809554 on Build GRCh38
rs772611737 Chr.16: 88809835-88809835 on Build GRCh38
rs140070863 Chr.16: 88809722-88809722 on Build GRCh38
rs764972083 Chr.16: 88809521-88809521 on Build GRCh38
rs957475910 Chr.16: 88809625-88809625 on Build GRCh38
rs569734625 Chr.16: 88809810-88809810 on Build GRCh38
rs931740936 Chr.16: 88809641-88809641 on Build GRCh38
rs28999113 Chr.16: 88809834-88809834 on Build GRCh38
rs913057078 Chr.16: 88809638-88809638 on Build GRCh38
rs748634790 Chr.16: 88809615-88809615 on Build GRCh38
rs752469016 Chr.16: 88809956-88809956 on Build GRCh38
rs760230688 Chr.16: 88810028-88810028 on Build GRCh38
rs549382452 Chr.16: 88809659-88809659 on Build GRCh38
rs749413936 Chr.16: 88809930-88809931 on Build GRCh38
rs769271336 Chr.16: 88809576-88809576 on Build GRCh38
rs746631133 Chr.16: 88810032-88810032 on Build GRCh38
rs1049298789 Chr.16: 88809745-88809745 on Build GRCh38
rs561477057 Chr.16: 88809519-88809519 on Build GRCh38
rs145607240 Chr.16: 88809797-88809797 on Build GRCh38
rs142472789 Chr.16: 88809830-88809830 on Build GRCh38
rs1018982587 Chr.16: 88809603-88809603 on Build GRCh38
rs8191495 Chr.16: 88809891-88809891 on Build GRCh38
rs990183908 Chr.16: 88809962-88809962 on Build GRCh38
rs774880744 Chr.16: 88810053-88810053 on Build GRCh38
rs771893788 Chr.16: 88810069-88810078 on Build GRCh38
rs745594160 Chr.16: 88810067-88810068 on Build GRCh38
rs1027745396 Chr.16: 88809906-88809906 on Build GRCh38
rs780365387 Chr.16: 88809563-88809563 on Build GRCh38
rs281860266 Chr.16: 88809793-88809793 on Build GRCh38
rs139444461 Chr.16: 88810042-88810042 on Build GRCh38
rs4695 Chr.16: 88809520-88809520 on Build GRCh38
rs747755586 Chr.16: 88809829-88809829 on Build GRCh38
rs764410865 Chr.16: 88809859-88809859 on Build GRCh38
rs981211482 Chr.16: 88809905-88809905 on Build GRCh38
rs753976270 Chr.16: 88810064-88810064 on Build GRCh38
rs763355047 Chr.16: 88809854-88809854 on Build GRCh38
rs879204584 Chr.16: 88810004-88810004 on Build GRCh38
rs367605558 Chr.16: 88809770-88809770 on Build GRCh38
rs377667293 Chr.16: 88809918-88809918 on Build GRCh38
rs374275550 Chr.16: 88809684-88809684 on Build GRCh38
rs777809251 Chr.16: 88809914-88809929 on Build GRCh38
rs774100663 Chr.16: 88809754-88809754 on Build GRCh38
rs781123799 Chr.16: 88809800-88809800 on Build GRCh38
rs930907433 Chr.16: 88809977-88809977 on Build GRCh38
rs767718095 Chr.16: 88809881-88809881 on Build GRCh38
rs555354866 Chr.16: 88809907-88809907 on Build GRCh38
rs1049331264 Chr.16: 88809997-88809997 on Build GRCh38
rs767663359 Chr.16: 88809936-88809936 on Build GRCh38
rs766909450 Chr.16: 88809759-88809759 on Build GRCh38
rs553748192 Chr.16: 88809999-88809999 on Build GRCh38
rs921715895 Chr.16: 88809640-88809640 on Build GRCh38
rs567519373 Chr.16: 88809687-88809687 on Build GRCh38
rs761438940 Chr.16: 88809758-88809758 on Build GRCh38
rs1020098230 Chr.16: 88809889-88809889 on Build GRCh38
rs374197629 Chr.16: 88810059-88810059 on Build GRCh38
rs745761689 Chr.16: 88809729-88809729 on Build GRCh38
rs774658385 Chr.16: 88810063-88810063 on Build GRCh38
rs775925348 Chr.16: 88809850-88809850 on Build GRCh38
rs758724286 Chr.16: 88809783-88809783 on Build GRCh38
rs550667813 Chr.16: 88809808-88809808 on Build GRCh38
rs369966180 Chr.16: 88809575-88809575 on Build GRCh38
rs560072301 Chr.16: 88809543-88809543 on Build GRCh38
rs749210314 Chr.16: 88809888-88809888 on Build GRCh38
rs769067195 Chr.16: 88809921-88809921 on Build GRCh38
rs777336565 Chr.16: 88810031-88810031 on Build GRCh38
rs757402931 Chr.16: 88809868-88809868 on Build GRCh38
rs771105725 Chr.16: 88809940-88809941 on Build GRCh38
rs778847710 Chr.16: 88809949-88809949 on Build GRCh38
rs750471263 Chr.16: 88809882-88809882 on Build GRCh38
rs751820457 Chr.16: 88809703-88809706 on Build GRCh38
rs8191498 Chr.16: 88809516-88809516 on Build GRCh38
rs750638200 Chr.16: 88809938-88809938 on Build GRCh38
rs780192617 Chr.16: 88809885-88809885 on Build GRCh38
rs781776802 Chr.16: 88809610-88809610 on Build GRCh38
rs141669365 Chr.16: 88809781-88809781 on Build GRCh38
rs121912681 Chr.16: 88809722-88809724 on Build GRCh38
rs774970490 Chr.16: 88809894-88809894 on Build GRCh38
rs756672917 Chr.16: 88809810-88809810 on Build GRCh38
rs776390101 Chr.16: 88809654-88809654 on Build GRCh38
rs773463923 Chr.16: 88809668-88809668 on Build GRCh38
rs577867349 Chr.16: 88810005-88810005 on Build GRCh38
rs766484349 Chr.16: 88809614-88809614 on Build GRCh38
rs777907885 Chr.16: 88809713-88809713 on Build GRCh38
rs1041573269 Chr.16: 88809864-88809864 on Build GRCh38
rs781486778 Chr.16: 88809713-88809714 on Build GRCh38
rs774591384 Chr.16: 88809930-88809930 on Build GRCh38
rs754262300 Chr.16: 88809567-88809567 on Build GRCh38
rs375131814 Chr.16: 88809782-88809782 on Build GRCh38
rs758634272 Chr.16: 88809700-88809700 on Build GRCh38
rs1003586349 Chr.16: 88809708-88809708 on Build GRCh38
rs989947379 Chr.16: 88809635-88809635 on Build GRCh38
rs181364821 Chr.16: 88809544-88809544 on Build GRCh38
rs1028927368 Chr.16: 88809616-88809616 on Build GRCh38
rs769454426 Chr.16: 88809666-88809666 on Build GRCh38
rs34926513 Chr.16: 88809908-88809909 on Build GRCh38
rs895364227 Chr.16: 88809561-88809561 on Build GRCh38
rs976381176 Chr.16: 88809942-88809942 on Build GRCh38
rs368883030 Chr.16: 88810055-88810055 on Build GRCh38
rs762170184 Chr.16: 88809934-88809934 on Build GRCh38
rs754983641 Chr.16: 88809948-88809948 on Build GRCh38
rs375180338 Chr.16: 88809696-88809696 on Build GRCh38
rs777265858 Chr.16: 88809769-88809769 on Build GRCh38
rs770492274 Chr.16: 88810041-88810041 on Build GRCh38
rs919583436 Chr.16: 88809976-88809976 on Build GRCh38
rs775317972 Chr.16: 88809667-88809667 on Build GRCh38
rs759652584 Chr.16: 88810073-88810073 on Build GRCh38
rs1035519732 Chr.16: 88809531-88809531 on Build GRCh38
rs370980256 Chr.16: 88809720-88809720 on Build GRCh38
rs764321075 Chr.16: 88809784-88809784 on Build GRCh38
rs373048958 Chr.16: 88810056-88810056 on Build GRCh38
rs375833985 Chr.16: 88809947-88809947 on Build GRCh38
rs1045406254 Chr.16: 88809517-88809517 on Build GRCh38
rs777817751 Chr.16: 88809649-88809649 on Build GRCh38
rs774096932 Chr.16: 88809818-88809818 on Build GRCh38
rs747919930 Chr.16: 88809897-88809897 on Build GRCh38
rs748342342 Chr.16: 88809577-88809577 on Build GRCh38
rs775204282 Chr.16: 88810073-88810082 on Build GRCh38
rs751125608 Chr.16: 88809677-88809677 on Build GRCh38
rs966060191 Chr.16: 88809611-88809611 on Build GRCh38
rs775409590 Chr.16: 88809734-88809734 on Build GRCh38
rs745324089 Chr.16: 88810043-88810043 on Build GRCh38
rs186437049 Chr.16: 88809993-88809993 on Build GRCh38
rs528396845 Chr.16: 88809652-88809652 on Build GRCh38
rs367973305 Chr.16: 88809846-88809846 on Build GRCh38
rs281860266 Chr.16: 88809793-88809793 on Build GRCh38
rs760165905 Chr.16: 88809842-88809842 on Build GRCh38
rs1005047521 Chr.16: 88809923-88809923 on Build GRCh38
rs760967219 Chr.16: 88810057-88810057 on Build GRCh38

View Full Product Details