Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145650939-145651426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
UniGene
Hs.452864
Gene Chromosome Location
Chr.4: 145619388-145660035 on Build GRCh38
Transcripts
NM_172250.2 XM_011531684.2 XM_011531685.2 XM_011531686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3760417 c.747G>A p.S249S Chr.4: 145651075-145651075 on Build GRCh38
4122568 c.773A>G p.D258G Chr.4: 145651101-145651101 on Build GRCh38
4896585 c.741T>G p.G247G Chr.4: 145651069-145651069 on Build GRCh38
4618397 c.746C>T p.S249L Chr.4: 145651074-145651074 on Build GRCh38
5520377 c.741T>A p.G247G Chr.4: 145651069-145651069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769722136 Chr.4: 145651088-145651088 on Build GRCh38
rs1036899492 Chr.4: 145651388-145651388 on Build GRCh38
rs757242348 Chr.4: 145651136-145651136 on Build GRCh38
rs978261211 Chr.4: 145651216-145651216 on Build GRCh38
rs576798223 Chr.4: 145651225-145651225 on Build GRCh38
rs773823789 Chr.4: 145651018-145651023 on Build GRCh38
rs187780119 Chr.4: 145650966-145650966 on Build GRCh38
rs539970676 Chr.4: 145651337-145651337 on Build GRCh38
rs201234437 Chr.4: 145651172-145651172 on Build GRCh38
rs752000155 Chr.4: 145651023-145651023 on Build GRCh38
rs759947234 Chr.4: 145651115-145651115 on Build GRCh38
rs753938949 Chr.4: 145651055-145651055 on Build GRCh38
rs975110848 Chr.4: 145651308-145651308 on Build GRCh38
rs1003941125 Chr.4: 145651031-145651031 on Build GRCh38
rs767934772 Chr.4: 145651123-145651123 on Build GRCh38
rs888293659 Chr.4: 145651017-145651017 on Build GRCh38
rs764675342 Chr.4: 145651131-145651131 on Build GRCh38
rs1033389087 Chr.4: 145651090-145651090 on Build GRCh38
rs536601590 Chr.4: 145651133-145651133 on Build GRCh38
rs761331762 Chr.4: 145651130-145651130 on Build GRCh38
rs545631797 Chr.4: 145651211-145651211 on Build GRCh38
rs748721023 Chr.4: 145651095-145651095 on Build GRCh38
rs1027955832 Chr.4: 145651389-145651389 on Build GRCh38
rs1003295681 Chr.4: 145651402-145651402 on Build GRCh38
rs1042658064 Chr.4: 145651314-145651314 on Build GRCh38
rs770636513 Chr.4: 145651103-145651103 on Build GRCh38
rs949694703 Chr.4: 145651276-145651276 on Build GRCh38
rs746853773 Chr.4: 145651074-145651074 on Build GRCh38
rs1033726836 Chr.4: 145651404-145651404 on Build GRCh38
rs996919253 Chr.4: 145651370-145651370 on Build GRCh38
rs781028629 Chr.4: 145651171-145651171 on Build GRCh38
rs765912031 Chr.4: 145651021-145651021 on Build GRCh38
rs765724008 Chr.4: 145651049-145651049 on Build GRCh38
rs11721510 Chr.4: 145650988-145650988 on Build GRCh38
rs72950836 Chr.4: 145651399-145651399 on Build GRCh38
rs138854691 Chr.4: 145651128-145651128 on Build GRCh38
rs758629317 Chr.4: 145651170-145651170 on Build GRCh38
rs894831249 Chr.4: 145651406-145651406 on Build GRCh38
rs559790918 Chr.4: 145651341-145651341 on Build GRCh38
rs773833728 Chr.4: 145651114-145651114 on Build GRCh38
rs545628307 Chr.4: 145651227-145651227 on Build GRCh38
rs765498620 Chr.4: 145651169-145651169 on Build GRCh38
rs72723819 Chr.4: 145651315-145651315 on Build GRCh38
rs748863136 Chr.4: 145651187-145651187 on Build GRCh38
rs777641906 Chr.4: 145651185-145651185 on Build GRCh38
rs756275016 Chr.4: 145651053-145651053 on Build GRCh38
rs892863115 Chr.4: 145651361-145651361 on Build GRCh38
rs11721553 Chr.4: 145651075-145651075 on Build GRCh38
rs757548934 Chr.4: 145651070-145651070 on Build GRCh38
rs778057240 Chr.4: 145651054-145651054 on Build GRCh38
rs531258230 Chr.4: 145651342-145651342 on Build GRCh38
rs558860546 Chr.4: 145651272-145651272 on Build GRCh38
rs748145761 Chr.4: 145651085-145651085 on Build GRCh38
rs888387475 Chr.4: 145651372-145651372 on Build GRCh38
rs778955528 Chr.4: 145651072-145651072 on Build GRCh38
rs1054220988 Chr.4: 145651339-145651339 on Build GRCh38
rs753187744 Chr.4: 145651050-145651050 on Build GRCh38
rs939821262 Chr.4: 145651384-145651384 on Build GRCh38
rs759994810 Chr.4: 145650995-145650995 on Build GRCh38
rs145018955 Chr.4: 145651089-145651089 on Build GRCh38
rs759767682 Chr.4: 145651048-145651048 on Build GRCh38
rs781124654 Chr.4: 145651084-145651084 on Build GRCh38

View Full Product Details