Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93793203-93793710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGI1
Entrez Gene ID
9211
Gene Name
leucine rich glioma inactivated 1
Gene Aliases
ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099
UniGene
Hs.533670
Gene Chromosome Location
Chr.10: 93757770-93798174 on Build GRCh38
Transcripts
NM_001308275.1 NM_001308276.1 NM_005097.3 NR_131777.1 XM_017016911.1 XM_017016912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3441736 c.742G>T p.E248* Chr.10: 93793254-93793254 on Build GRCh38
3441737 c.805G>A p.E269K Chr.10: 93793317-93793317 on Build GRCh38
4697844 c.804G>A p.V268V Chr.10: 93793316-93793316 on Build GRCh38
312487 c.795G>A p.W265* Chr.10: 93793307-93793307 on Build GRCh38
1970202 c.709T>C p.L237L Chr.10: 93793221-93793221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548803120 Chr.10: 93793277-93793277 on Build GRCh38
rs797044997 Chr.10: 93793270-93793270 on Build GRCh38
rs754980689 Chr.10: 93793234-93793234 on Build GRCh38
rs910029404 Chr.10: 93793647-93793647 on Build GRCh38
rs552777218 Chr.10: 93793293-93793293 on Build GRCh38
rs750249658 Chr.10: 93793330-93793330 on Build GRCh38
rs755068491 Chr.10: 93793349-93793349 on Build GRCh38
rs781404129 Chr.10: 93793250-93793250 on Build GRCh38
rs780241372 Chr.10: 93793228-93793228 on Build GRCh38
rs1009414757 Chr.10: 93793511-93793511 on Build GRCh38
rs937646863 Chr.10: 93793405-93793405 on Build GRCh38
rs961648156 Chr.10: 93793684-93793684 on Build GRCh38
rs781205119 Chr.10: 93793353-93793353 on Build GRCh38
rs373818254 Chr.10: 93793396-93793396 on Build GRCh38
rs185474566 Chr.10: 93793545-93793545 on Build GRCh38
rs777252300 Chr.10: 93793501-93793501 on Build GRCh38
rs535138541 Chr.10: 93793577-93793577 on Build GRCh38
rs748315756 Chr.10: 93793253-93793253 on Build GRCh38
rs906016145 Chr.10: 93793399-93793399 on Build GRCh38
rs766797914 Chr.10: 93793343-93793343 on Build GRCh38
rs909649461 Chr.10: 93793379-93793379 on Build GRCh38
rs748409467 Chr.10: 93793358-93793358 on Build GRCh38
rs759132595 Chr.10: 93793313-93793313 on Build GRCh38
rs886047486 Chr.10: 93793361-93793361 on Build GRCh38
rs752003436 Chr.10: 93793347-93793347 on Build GRCh38
rs982418045 Chr.10: 93793366-93793366 on Build GRCh38
rs888991188 Chr.10: 93793443-93793443 on Build GRCh38
rs377206644 Chr.10: 93793337-93793337 on Build GRCh38
rs911013524 Chr.10: 93793243-93793243 on Build GRCh38
rs951255631 Chr.10: 93793683-93793683 on Build GRCh38
rs757949428 Chr.10: 93793481-93793481 on Build GRCh38
rs746376861 Chr.10: 93793553-93793553 on Build GRCh38
rs576944198 Chr.10: 93793505-93793505 on Build GRCh38
rs749031343 Chr.10: 93793380-93793380 on Build GRCh38
rs941064994 Chr.10: 93793394-93793394 on Build GRCh38
rs865793574 Chr.10: 93793340-93793340 on Build GRCh38
rs551265218 Chr.10: 93793484-93793484 on Build GRCh38
rs146425212 Chr.10: 93793229-93793229 on Build GRCh38
rs777679645 Chr.10: 93793377-93793377 on Build GRCh38
rs140752487 Chr.10: 93793324-93793324 on Build GRCh38
rs780606965 Chr.10: 93793380-93793381 on Build GRCh38
rs552841524 Chr.10: 93793521-93793521 on Build GRCh38
rs893735567 Chr.10: 93793508-93793508 on Build GRCh38
rs368264015 Chr.10: 93793268-93793268 on Build GRCh38
rs571391768 Chr.10: 93793341-93793341 on Build GRCh38
rs976727922 Chr.10: 93793634-93793634 on Build GRCh38
rs372978335 Chr.10: 93793280-93793280 on Build GRCh38
rs146600209 Chr.10: 93793459-93793459 on Build GRCh38
rs959119806 Chr.10: 93793549-93793549 on Build GRCh38
rs774280395 Chr.10: 93793304-93793304 on Build GRCh38
rs756267808 Chr.10: 93793360-93793360 on Build GRCh38
rs568626175 Chr.10: 93793404-93793404 on Build GRCh38
rs770758936 Chr.10: 93793283-93793283 on Build GRCh38
rs760551519 Chr.10: 93793466-93793466 on Build GRCh38
rs745760611 Chr.10: 93793400-93793400 on Build GRCh38
rs770702247 Chr.10: 93793390-93793390 on Build GRCh38
rs760376686 Chr.10: 93793329-93793329 on Build GRCh38
rs544843166 Chr.10: 93793514-93793514 on Build GRCh38
rs199916185 Chr.10: 93793256-93793256 on Build GRCh38
rs373248621 Chr.10: 93793398-93793398 on Build GRCh38
rs367701497 Chr.10: 93793520-93793520 on Build GRCh38
rs555159644 Chr.10: 93793630-93793630 on Build GRCh38

View Full Product Details