Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66638989-66639529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD7
Entrez Gene ID
154881
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
CLN14, EPM3
UniGene
Hs.546627
Gene Chromosome Location
Chr.7: 66628881-66643229 on Build GRCh38
Transcripts
NM_001167961.2 NM_153033.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4558019 c.751G>A p.D251N Chr.7: 66639113-66639113 on Build GRCh38
4616644 c.735G>T p.L245L Chr.7: 66639097-66639097 on Build GRCh38
80044 c.816G>C p.V272V Chr.7: 66639178-66639178 on Build GRCh38
4502976 c.627C>T p.S209S Chr.7: 66638989-66638989 on Build GRCh38
6692117 c.759G>A p.S253S Chr.7: 66639121-66639121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927709940 Chr.7: 66639169-66639169 on Build GRCh38
rs369862253 Chr.7: 66639027-66639027 on Build GRCh38
rs748183090 Chr.7: 66639225-66639225 on Build GRCh38
rs989382981 Chr.7: 66639106-66639106 on Build GRCh38
rs1023493281 Chr.7: 66639454-66639454 on Build GRCh38
rs932179746 Chr.7: 66639377-66639377 on Build GRCh38
rs201990302 Chr.7: 66639061-66639061 on Build GRCh38
rs763830869 Chr.7: 66639152-66639152 on Build GRCh38
rs940752551 Chr.7: 66639349-66639349 on Build GRCh38
rs374256335 Chr.7: 66639112-66639112 on Build GRCh38
rs758401521 Chr.7: 66639284-66639284 on Build GRCh38
rs907869743 Chr.7: 66639346-66639346 on Build GRCh38
rs368680616 Chr.7: 66639154-66639154 on Build GRCh38
rs756747620 Chr.7: 66639256-66639256 on Build GRCh38
rs773953605 Chr.7: 66639236-66639236 on Build GRCh38
rs367591875 Chr.7: 66639223-66639223 on Build GRCh38
rs141191660 Chr.7: 66639189-66639189 on Build GRCh38
rs116630203 Chr.7: 66639241-66639241 on Build GRCh38
rs774927044 Chr.7: 66639242-66639242 on Build GRCh38
rs766720725 Chr.7: 66639125-66639125 on Build GRCh38
rs749877863 Chr.7: 66639266-66639266 on Build GRCh38
rs1014919686 Chr.7: 66639500-66639500 on Build GRCh38
rs763849444 Chr.7: 66639248-66639248 on Build GRCh38
rs200415747 Chr.7: 66639155-66639155 on Build GRCh38
rs960570960 Chr.7: 66639222-66639222 on Build GRCh38
rs113446781 Chr.7: 66639383-66639383 on Build GRCh38
rs145063728 Chr.7: 66639197-66639197 on Build GRCh38
rs761940050 Chr.7: 66639120-66639120 on Build GRCh38
rs951958149 Chr.7: 66639263-66639263 on Build GRCh38
rs969666860 Chr.7: 66639476-66639476 on Build GRCh38
rs541759714 Chr.7: 66639382-66639382 on Build GRCh38
rs923444735 Chr.7: 66639365-66639365 on Build GRCh38
rs117194263 Chr.7: 66639016-66639016 on Build GRCh38
rs766332054 Chr.7: 66639175-66639175 on Build GRCh38
rs777050711 Chr.7: 66639133-66639133 on Build GRCh38
rs746774522 Chr.7: 66639196-66639196 on Build GRCh38
rs976607982 Chr.7: 66639351-66639351 on Build GRCh38
rs922098847 Chr.7: 66639336-66639336 on Build GRCh38
rs763260837 Chr.7: 66639123-66639123 on Build GRCh38
rs751338237 Chr.7: 66639176-66639176 on Build GRCh38
rs796052689 Chr.7: 66639066-66639066 on Build GRCh38
rs958211575 Chr.7: 66639083-66639083 on Build GRCh38
rs768574038 Chr.7: 66639203-66639203 on Build GRCh38
rs932229206 Chr.7: 66639118-66639118 on Build GRCh38
rs372150992 Chr.7: 66639049-66639049 on Build GRCh38
rs149123496 Chr.7: 66639334-66639334 on Build GRCh38
rs751953589 Chr.7: 66639136-66639136 on Build GRCh38
rs1052877177 Chr.7: 66639431-66639431 on Build GRCh38
rs151151235 Chr.7: 66639453-66639453 on Build GRCh38
rs556805454 Chr.7: 66639076-66639076 on Build GRCh38
rs77341088 Chr.7: 66639448-66639448 on Build GRCh38
rs1003311931 Chr.7: 66639477-66639477 on Build GRCh38
rs947639717 Chr.7: 66639391-66639391 on Build GRCh38
rs886043127 Chr.7: 66639146-66639146 on Build GRCh38
rs779871558 Chr.7: 66639194-66639194 on Build GRCh38
rs776525362 Chr.7: 66639438-66639438 on Build GRCh38
rs759961871 Chr.7: 66639247-66639247 on Build GRCh38
rs374777888 Chr.7: 66639098-66639098 on Build GRCh38
rs778718424 Chr.7: 66639174-66639174 on Build GRCh38
rs375872253 Chr.7: 66639240-66639240 on Build GRCh38
rs760837610 Chr.7: 66639111-66639111 on Build GRCh38
rs1035207417 Chr.7: 66639443-66639443 on Build GRCh38
rs985121432 Chr.7: 66639469-66639469 on Build GRCh38
rs753471697 Chr.7: 66639254-66639254 on Build GRCh38
rs200652879 Chr.7: 66639116-66639116 on Build GRCh38
rs764754359 Chr.7: 66639260-66639260 on Build GRCh38
rs387907261 Chr.7: 66639180-66639180 on Build GRCh38
rs941670086 Chr.7: 66639186-66639194 on Build GRCh38
rs886062414 Chr.7: 66639287-66639287 on Build GRCh38
rs890525029 Chr.7: 66639392-66639392 on Build GRCh38
rs755875407 Chr.7: 66639143-66639143 on Build GRCh38
rs893768954 Chr.7: 66639432-66639432 on Build GRCh38
rs771832195 Chr.7: 66639085-66639085 on Build GRCh38
rs769596185 Chr.7: 66639506-66639506 on Build GRCh38
rs146540578 Chr.7: 66639121-66639121 on Build GRCh38
rs768924233 Chr.7: 66639198-66639198 on Build GRCh38
rs999586476 Chr.7: 66639415-66639415 on Build GRCh38
rs776777218 Chr.7: 66639214-66639214 on Build GRCh38
rs1037925062 Chr.7: 66639295-66639295 on Build GRCh38

View Full Product Details