Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96120350-96120949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A13
Entrez Gene ID
10165
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
UniGene
Hs.489190
Gene Chromosome Location
Chr.7: 96120220-96322147 on Build GRCh38
Transcripts
NM_001160210.1 NM_014251.2 NR_027662.1 XM_006715831.3 XM_017011663.1 XM_017011664.1 XM_017011665.1

SNP Detail

SNP ID Chromosome Location
rs559778122 Chr.7: 96120688-96120688 on Build GRCh38
rs377554502 Chr.7: 96120811-96120811 on Build GRCh38
rs376925315 Chr.7: 96120600-96120600 on Build GRCh38
rs182837763 Chr.7: 96120710-96120710 on Build GRCh38
rs990190192 Chr.7: 96120778-96120778 on Build GRCh38
rs963726789 Chr.7: 96120790-96120790 on Build GRCh38
rs574148664 Chr.7: 96120604-96120604 on Build GRCh38
rs898770101 Chr.7: 96120603-96120603 on Build GRCh38
rs886062525 Chr.7: 96120861-96120861 on Build GRCh38
rs924521499 Chr.7: 96120712-96120712 on Build GRCh38
rs924580530 Chr.7: 96120392-96120392 on Build GRCh38
rs373153995 Chr.7: 96120462-96120462 on Build GRCh38
rs993746651 Chr.7: 96120389-96120389 on Build GRCh38
rs954232609 Chr.7: 96120658-96120658 on Build GRCh38
rs1018706685 Chr.7: 96120693-96120693 on Build GRCh38
rs983911109 Chr.7: 96120434-96120434 on Build GRCh38
rs574119069 Chr.7: 96120399-96120399 on Build GRCh38
rs769226693 Chr.7: 96120388-96120388 on Build GRCh38
rs187381119 Chr.7: 96120765-96120765 on Build GRCh38
rs1017988082 Chr.7: 96120782-96120782 on Build GRCh38
rs761315073 Chr.7: 96120571-96120571 on Build GRCh38
rs368958557 Chr.7: 96120411-96120411 on Build GRCh38
rs538235683 Chr.7: 96120458-96120458 on Build GRCh38
rs770563944 Chr.7: 96120441-96120441 on Build GRCh38
rs192117485 Chr.7: 96120794-96120794 on Build GRCh38
rs887970622 Chr.7: 96120733-96120733 on Build GRCh38
rs1028304408 Chr.7: 96120728-96120728 on Build GRCh38
rs766684721 Chr.7: 96120735-96120735 on Build GRCh38
rs771843737 Chr.7: 96120918-96120918 on Build GRCh38
rs925831789 Chr.7: 96120898-96120898 on Build GRCh38
rs558607926 Chr.7: 96120470-96120470 on Build GRCh38
rs986866433 Chr.7: 96120659-96120659 on Build GRCh38
rs935854981 Chr.7: 96120909-96120909 on Build GRCh38
rs759111517 Chr.7: 96120464-96120464 on Build GRCh38
rs965839734 Chr.7: 96120699-96120699 on Build GRCh38
rs1045800989 Chr.7: 96120708-96120708 on Build GRCh38
rs967007036 Chr.7: 96120868-96120868 on Build GRCh38
rs781554158 Chr.7: 96120813-96120813 on Build GRCh38
rs995908248 Chr.7: 96120616-96120616 on Build GRCh38
rs34219676 Chr.7: 96120889-96120890 on Build GRCh38
rs575375923 Chr.7: 96120569-96120569 on Build GRCh38
rs906980053 Chr.7: 96120905-96120905 on Build GRCh38
rs948426527 Chr.7: 96120833-96120833 on Build GRCh38
rs925829411 Chr.7: 96120594-96120594 on Build GRCh38
rs373207040 Chr.7: 96120882-96120882 on Build GRCh38
rs946008706 Chr.7: 96120580-96120580 on Build GRCh38
rs796781003 Chr.7: 96120780-96120780 on Build GRCh38
rs767379676 Chr.7: 96120588-96120588 on Build GRCh38
rs1014265735 Chr.7: 96120657-96120657 on Build GRCh38
rs886062524 Chr.7: 96120684-96120684 on Build GRCh38
rs761805247 Chr.7: 96120624-96120624 on Build GRCh38
rs758761551 Chr.7: 96120713-96120713 on Build GRCh38
rs544333452 Chr.7: 96120587-96120587 on Build GRCh38
rs530869704 Chr.7: 96120881-96120881 on Build GRCh38
rs550661000 Chr.7: 96120912-96120912 on Build GRCh38
rs147716687 Chr.7: 96120621-96120621 on Build GRCh38
rs765351576 Chr.7: 96120692-96120692 on Build GRCh38
rs886062523 Chr.7: 96120674-96120674 on Build GRCh38
rs532132551 Chr.7: 96120507-96120512 on Build GRCh38

View Full Product Details