Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112454542-112454694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 XM_006719526.2 XM_011538613.2 XM_017019722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338919 c.560T>A p.F187Y Chr.12: 112454598-112454598 on Build GRCh38
6951923 c.601C>T p.P201S Chr.12: 112454639-112454639 on Build GRCh38
1723261 c.540C>T p.D180D Chr.12: 112454578-112454578 on Build GRCh38
5905919 c.602C>T p.P201L Chr.12: 112454640-112454640 on Build GRCh38
5413312 c.537C>T p.Y179Y Chr.12: 112454575-112454575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143433437 Chr.12: 112454594-112454594 on Build GRCh38
rs727503381 Chr.12: 112454636-112454636 on Build GRCh38
rs778211349 Chr.12: 112454632-112454632 on Build GRCh38
rs141015445 Chr.12: 112454575-112454575 on Build GRCh38
rs753107691 Chr.12: 112454623-112454623 on Build GRCh38
rs753269427 Chr.12: 112454578-112454578 on Build GRCh38
rs778919108 Chr.12: 112454668-112454668 on Build GRCh38
rs757304894 Chr.12: 112454659-112454659 on Build GRCh38
rs370763866 Chr.12: 112454610-112454610 on Build GRCh38
rs764663951 Chr.12: 112454595-112454595 on Build GRCh38
rs76982592 Chr.12: 112454629-112454629 on Build GRCh38
rs79068130 Chr.12: 112454603-112454603 on Build GRCh38
rs78376169 Chr.12: 112454654-112454654 on Build GRCh38
rs864622409 Chr.12: 112454662-112454662 on Build GRCh38
rs745331641 Chr.12: 112454670-112454670 on Build GRCh38
rs200920312 Chr.12: 112454596-112454596 on Build GRCh38

View Full Product Details