Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72343414-72343963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1

SNP Detail

SNP ID Chromosome Location
rs935987029 Chr.15: 72343513-72343513 on Build GRCh38
rs989275703 Chr.15: 72343505-72343505 on Build GRCh38
rs34300017 Chr.15: 72343895-72343895 on Build GRCh38
rs560068788 Chr.15: 72343808-72343808 on Build GRCh38
rs1033099973 Chr.15: 72343681-72343681 on Build GRCh38
rs1033088636 Chr.15: 72343750-72343750 on Build GRCh38
rs112626309 Chr.15: 72343641-72343641 on Build GRCh38
rs763246017 Chr.15: 72343809-72343809 on Build GRCh38
rs778510381 Chr.15: 72343539-72343539 on Build GRCh38
rs886051461 Chr.15: 72343877-72343877 on Build GRCh38
rs1046379119 Chr.15: 72343671-72343671 on Build GRCh38
rs926032419 Chr.15: 72343503-72343503 on Build GRCh38
rs1019249372 Chr.15: 72343791-72343791 on Build GRCh38
rs76075374 Chr.15: 72343502-72343502 on Build GRCh38
rs940337374 Chr.15: 72343612-72343612 on Build GRCh38
rs1054432 Chr.15: 72343894-72343894 on Build GRCh38
rs967502009 Chr.15: 72343729-72343729 on Build GRCh38
rs957471664 Chr.15: 72343754-72343754 on Build GRCh38
rs965759418 Chr.15: 72343718-72343718 on Build GRCh38
rs760592453 Chr.15: 72343697-72343697 on Build GRCh38
rs770433545 Chr.15: 72343508-72343508 on Build GRCh38
rs527803610 Chr.15: 72343921-72343921 on Build GRCh38
rs922739871 Chr.15: 72343453-72343453 on Build GRCh38
rs930392742 Chr.15: 72343637-72343637 on Build GRCh38
rs998816698 Chr.15: 72343720-72343720 on Build GRCh38
rs747982224 Chr.15: 72343446-72343446 on Build GRCh38
rs560253606 Chr.15: 72343640-72343640 on Build GRCh38
rs139968433 Chr.15: 72343759-72343759 on Build GRCh38
rs563352066 Chr.15: 72343857-72343857 on Build GRCh38
rs987425297 Chr.15: 72343766-72343766 on Build GRCh38
rs768562754 Chr.15: 72343702-72343702 on Build GRCh38
rs1036084404 Chr.15: 72343622-72343622 on Build GRCh38
rs920305614 Chr.15: 72343636-72343636 on Build GRCh38
rs555620598 Chr.15: 72343653-72343653 on Build GRCh38
rs35949555 Chr.15: 72343447-72343447 on Build GRCh38
rs766460302 Chr.15: 72343829-72343829 on Build GRCh38
rs375308096 Chr.15: 72343938-72343938 on Build GRCh38
rs950232590 Chr.15: 72343937-72343937 on Build GRCh38
rs920253133 Chr.15: 72343846-72343846 on Build GRCh38
rs11629508 Chr.15: 72343488-72343488 on Build GRCh38
rs188399769 Chr.15: 72343722-72343722 on Build GRCh38
rs751716928 Chr.15: 72343903-72343903 on Build GRCh38
rs545085202 Chr.15: 72343784-72343784 on Build GRCh38
rs60288568 Chr.15: 72343528-72343528 on Build GRCh38
rs911586170 Chr.15: 72343563-72343563 on Build GRCh38
rs184059814 Chr.15: 72343710-72343710 on Build GRCh38
rs3087652 Chr.15: 72343562-72343562 on Build GRCh38
rs775453371 Chr.15: 72343674-72343674 on Build GRCh38
rs550241078 Chr.15: 72343467-72343467 on Build GRCh38
rs1045661428 Chr.15: 72343589-72343589 on Build GRCh38
rs916097934 Chr.15: 72343928-72343928 on Build GRCh38
rs1028855564 Chr.15: 72343745-72343745 on Build GRCh38
rs573845046 Chr.15: 72343677-72343677 on Build GRCh38
rs868362186 Chr.15: 72343924-72343924 on Build GRCh38

View Full Product Details