Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 63059001-63059525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTPA
Entrez Gene ID
7274
Gene Name
alpha tocopherol transfer protein
Gene Aliases
ATTP, AVED, TTP1, alphaTTP
UniGene
Hs.69049
Gene Chromosome Location
Chr.8: 63058409-63086523 on Build GRCh38
Transcripts
NM_000370.3 XM_006716468.3

SNP Detail

SNP ID Chromosome Location
rs867141767 Chr.8: 63059157-63059157 on Build GRCh38
rs554586373 Chr.8: 63059253-63059253 on Build GRCh38
rs565918859 Chr.8: 63059190-63059190 on Build GRCh38
rs907076427 Chr.8: 63059035-63059035 on Build GRCh38
rs910414252 Chr.8: 63059180-63059180 on Build GRCh38
rs895587319 Chr.8: 63059231-63059231 on Build GRCh38
rs902560154 Chr.8: 63059459-63059459 on Build GRCh38
rs1055182336 Chr.8: 63059072-63059072 on Build GRCh38
rs918284042 Chr.8: 63059303-63059304 on Build GRCh38
rs560197099 Chr.8: 63059401-63059401 on Build GRCh38
rs755311741 Chr.8: 63059156-63059157 on Build GRCh38
rs963275610 Chr.8: 63059284-63059284 on Build GRCh38
rs902762340 Chr.8: 63059160-63059160 on Build GRCh38
rs956081768 Chr.8: 63059147-63059147 on Build GRCh38
rs543160605 Chr.8: 63059462-63059462 on Build GRCh38
rs1010886177 Chr.8: 63059131-63059131 on Build GRCh38
rs941921823 Chr.8: 63059186-63059186 on Build GRCh38
rs576236825 Chr.8: 63059275-63059275 on Build GRCh38
rs939425475 Chr.8: 63059039-63059039 on Build GRCh38
rs1000854271 Chr.8: 63059243-63059243 on Build GRCh38
rs536415036 Chr.8: 63059241-63059241 on Build GRCh38
rs553883010 Chr.8: 63059071-63059071 on Build GRCh38
rs145257038 Chr.8: 63059312-63059312 on Build GRCh38
rs869310085 Chr.8: 63059044-63059045 on Build GRCh38
rs867977454 Chr.8: 63059028-63059028 on Build GRCh38
rs999722930 Chr.8: 63059463-63059463 on Build GRCh38
rs928932514 Chr.8: 63059307-63059307 on Build GRCh38
rs1053743092 Chr.8: 63059404-63059404 on Build GRCh38
rs867160086 Chr.8: 63059027-63059027 on Build GRCh38
rs936191833 Chr.8: 63059400-63059400 on Build GRCh38
rs9792280 Chr.8: 63059170-63059170 on Build GRCh38
rs892057788 Chr.8: 63059428-63059428 on Build GRCh38
rs190343442 Chr.8: 63059279-63059279 on Build GRCh38
rs1022268395 Chr.8: 63059150-63059150 on Build GRCh38
rs998145520 Chr.8: 63059267-63059267 on Build GRCh38
rs918483847 Chr.8: 63059025-63059025 on Build GRCh38
rs796706457 Chr.8: 63059259-63059259 on Build GRCh38
rs1029670787 Chr.8: 63059272-63059272 on Build GRCh38
rs928963792 Chr.8: 63059209-63059209 on Build GRCh38
rs895223494 Chr.8: 63059078-63059078 on Build GRCh38
rs973692910 Chr.8: 63059291-63059291 on Build GRCh38
rs973382880 Chr.8: 63059188-63059188 on Build GRCh38
rs904683160 Chr.8: 63059384-63059384 on Build GRCh38
rs976798137 Chr.8: 63059165-63059165 on Build GRCh38
rs1044193442 Chr.8: 63059358-63059358 on Build GRCh38
rs919184071 Chr.8: 63059304-63059304 on Build GRCh38
rs1029781715 Chr.8: 63059194-63059194 on Build GRCh38
rs12375357 Chr.8: 63059240-63059240 on Build GRCh38
rs1056054531 Chr.8: 63059375-63059375 on Build GRCh38
rs528499321 Chr.8: 63059319-63059319 on Build GRCh38
rs544287656 Chr.8: 63059276-63059276 on Build GRCh38
rs929890223 Chr.8: 63059026-63059026 on Build GRCh38
rs916948527 Chr.8: 63059325-63059325 on Build GRCh38
rs868357451 Chr.8: 63059156-63059156 on Build GRCh38
rs1029711550 Chr.8: 63059166-63059166 on Build GRCh38
rs1043690788 Chr.8: 63059437-63059437 on Build GRCh38
rs1056862669 Chr.8: 63059217-63059217 on Build GRCh38
rs200592548 Chr.8: 63059231-63059232 on Build GRCh38
rs986037992 Chr.8: 63059234-63059234 on Build GRCh38
rs527894836 Chr.8: 63059432-63059432 on Build GRCh38
rs948837174 Chr.8: 63059236-63059236 on Build GRCh38
rs1045576929 Chr.8: 63059032-63059032 on Build GRCh38
rs954519794 Chr.8: 63059167-63059167 on Build GRCh38
rs545125844 Chr.8: 63059328-63059328 on Build GRCh38

View Full Product Details