Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44408014-44408446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNF4A
Entrez Gene ID
3172
Gene Name
hepatocyte nuclear factor 4 alpha
Gene Aliases
FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14
UniGene
Hs.116462
Gene Chromosome Location
Chr.20: 44355801-44432845 on Build GRCh38
Transcripts
NM_000457.4 NM_001030003.2 NM_001030004.2 NM_001258355.1 NM_001287182.1 NM_001287183.1 NM_001287184.1 NM_175914.4 NM_178849.2 NM_178850.2 XM_005260407.3 NR_037419.1

Gene Symbol
MIR3646
Entrez Gene ID
100500813
Gene Name
microRNA 3646
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.20: 44408120-44408203 on Build GRCh38
Transcripts
NM_000457.4 NM_001030003.2 NM_001030004.2 NM_001258355.1 NM_001287182.1 NM_001287183.1 NM_001287184.1 NM_175914.4 NM_178849.2 NM_178850.2 XM_005260407.3 NR_037419.1

SNP Detail

SNP ID Chromosome Location
rs1020339709 Chr.20: 44408042-44408042 on Build GRCh38
rs11574730 Chr.20: 44408373-44408373 on Build GRCh38
rs932153460 Chr.20: 44408275-44408275 on Build GRCh38
rs8122476 Chr.20: 44408051-44408051 on Build GRCh38
rs980253035 Chr.20: 44408050-44408050 on Build GRCh38
rs996080641 Chr.20: 44408347-44408347 on Build GRCh38
rs1034210641 Chr.20: 44408054-44408054 on Build GRCh38
rs900410439 Chr.20: 44408321-44408321 on Build GRCh38
rs746206145 Chr.20: 44408072-44408072 on Build GRCh38
rs886405649 Chr.20: 44408180-44408180 on Build GRCh38
rs146092727 Chr.20: 44408123-44408123 on Build GRCh38
rs1047811712 Chr.20: 44408168-44408168 on Build GRCh38
rs974276600 Chr.20: 44408116-44408116 on Build GRCh38
rs577531393 Chr.20: 44408260-44408260 on Build GRCh38
rs907850014 Chr.20: 44408162-44408162 on Build GRCh38
rs951347833 Chr.20: 44408148-44408148 on Build GRCh38
rs537699859 Chr.20: 44408124-44408124 on Build GRCh38
rs559988739 Chr.20: 44408374-44408374 on Build GRCh38
rs190953299 Chr.20: 44408394-44408394 on Build GRCh38
rs966128643 Chr.20: 44408170-44408172 on Build GRCh38
rs746063392 Chr.20: 44408204-44408204 on Build GRCh38
rs760699353 Chr.20: 44408094-44408094 on Build GRCh38
rs555280096 Chr.20: 44408065-44408065 on Build GRCh38
rs958527821 Chr.20: 44408087-44408087 on Build GRCh38
rs1002602398 Chr.20: 44408052-44408052 on Build GRCh38
rs914839940 Chr.20: 44408318-44408318 on Build GRCh38
rs1054996862 Chr.20: 44408311-44408311 on Build GRCh38
rs140125029 Chr.20: 44408237-44408237 on Build GRCh38
rs1026913462 Chr.20: 44408133-44408133 on Build GRCh38
rs983490840 Chr.20: 44408157-44408157 on Build GRCh38

View Full Product Details