Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
Chromosome Location
Chr.3: 37862152 - 37984469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008392.1 NP_001008393.1 6 - 7 543 81
NM_005808.2 NP_005799.2 5 - 6 510 81
XM_017005519.1 XP_016861008.1 6 - 7 569 81
XM_017005520.1 XP_016861009.1 6 - 7 1887 81
GenBank mRNA AJ575644.1 - 5 - 6 510 81
AJ575645.1 - 6 - 7 543 81
AY279532.1 - 5 - 6 488 81
AY364238.1 - 5 - 6 488 81
D88153.1 - 6 - 7 860 81

View Full Product Details