Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40797882-40798431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A15
Entrez Gene ID
10166
Gene Name
solute carrier family 25 member 15
Gene Aliases
D13S327, HHH, ORC1, ORNT1
UniGene
Hs.646645
Gene Chromosome Location
Chr.13: 40789411-40812460 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Gene Symbol
TPTE2P5
Entrez Gene ID
100616668
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5
Gene Aliases
-
UniGene
Hs.660718
Gene Chromosome Location
Chr.13: 40796985-40921750 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

SNP Detail

SNP ID Chromosome Location
rs775436187 Chr.13: 40797993-40797993 on Build GRCh38
rs1013321987 Chr.13: 40798335-40798335 on Build GRCh38
rs898244586 Chr.13: 40798344-40798347 on Build GRCh38
rs187722549 Chr.13: 40798214-40798214 on Build GRCh38
rs1037918597 Chr.13: 40798202-40798202 on Build GRCh38
rs993747247 Chr.13: 40798246-40798246 on Build GRCh38
rs781078983 Chr.13: 40798279-40798280 on Build GRCh38
rs538532358 Chr.13: 40797911-40797911 on Build GRCh38
rs138800361 Chr.13: 40798000-40798000 on Build GRCh38
rs889094092 Chr.13: 40798157-40798157 on Build GRCh38
rs917569965 Chr.13: 40798002-40798002 on Build GRCh38
rs1052274839 Chr.13: 40798082-40798082 on Build GRCh38
rs995604189 Chr.13: 40798395-40798395 on Build GRCh38
rs777109424 Chr.13: 40797936-40797936 on Build GRCh38
rs774469251 Chr.13: 40798188-40798188 on Build GRCh38
rs969649399 Chr.13: 40798031-40798031 on Build GRCh38
rs1025316010 Chr.13: 40797996-40797996 on Build GRCh38
rs896013952 Chr.13: 40798331-40798331 on Build GRCh38
rs9603791 Chr.13: 40797964-40797964 on Build GRCh38
rs558149619 Chr.13: 40797926-40797926 on Build GRCh38
rs573503487 Chr.13: 40798394-40798394 on Build GRCh38
rs992359633 Chr.13: 40798001-40798001 on Build GRCh38
rs968061905 Chr.13: 40798143-40798143 on Build GRCh38
rs545342127 Chr.13: 40798399-40798399 on Build GRCh38
rs1051429170 Chr.13: 40798255-40798255 on Build GRCh38
rs912403141 Chr.13: 40798292-40798292 on Build GRCh38
rs58375222 Chr.13: 40798280-40798281 on Build GRCh38
rs573787530 Chr.13: 40798081-40798081 on Build GRCh38
rs939801614 Chr.13: 40798322-40798322 on Build GRCh38
rs559500402 Chr.13: 40798232-40798232 on Build GRCh38
rs988186566 Chr.13: 40798072-40798072 on Build GRCh38
rs969487014 Chr.13: 40798030-40798030 on Build GRCh38
rs1002813282 Chr.13: 40798058-40798058 on Build GRCh38
rs933417073 Chr.13: 40798105-40798105 on Build GRCh38
rs746130260 Chr.13: 40797944-40797944 on Build GRCh38
rs913770289 Chr.13: 40798092-40798092 on Build GRCh38
rs963491683 Chr.13: 40797988-40797988 on Build GRCh38
rs550314422 Chr.13: 40798140-40798140 on Build GRCh38
rs373728022 Chr.13: 40797935-40797935 on Build GRCh38
rs386378913 Chr.13: 40798294-40798295 on Build GRCh38
rs921211868 Chr.13: 40798221-40798221 on Build GRCh38
rs776698069 Chr.13: 40797907-40797907 on Build GRCh38
rs890925346 Chr.13: 40797910-40797910 on Build GRCh38
rs564905202 Chr.13: 40798400-40798400 on Build GRCh38
rs1026440968 Chr.13: 40798327-40798327 on Build GRCh38
rs183039437 Chr.13: 40798060-40798060 on Build GRCh38
rs397763536 Chr.13: 40798295-40798296 on Build GRCh38
rs913952346 Chr.13: 40798106-40798106 on Build GRCh38
rs5803069 Chr.13: 40798295-40798295 on Build GRCh38
rs925207914 Chr.13: 40798068-40798068 on Build GRCh38
rs78565724 Chr.13: 40798279-40798279 on Build GRCh38

View Full Product Details