Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 31058420-31058574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
UniGene
Hs.632050
Gene Chromosome Location
Chr.8: 31033262-31173761 on Build GRCh38
Transcripts
NM_000553.4 XM_011544639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7397942 c.95A>G p.K32R Chr.8: 31058542-31058542 on Build GRCh38
750650 c.96G>T p.K32N Chr.8: 31058543-31058543 on Build GRCh38
3731362 c.8delA p.K5fs*15 Chr.8: 31058455-31058455 on Build GRCh38
1099217 c.38G>A p.R13Q Chr.8: 31058485-31058485 on Build GRCh38
2958600 c.33G>A p.Q11Q Chr.8: 31058480-31058480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756255418 Chr.8: 31058480-31058480 on Build GRCh38
rs150524008 Chr.8: 31058522-31058522 on Build GRCh38
rs755891046 Chr.8: 31058543-31058543 on Build GRCh38
rs878854133 Chr.8: 31058462-31058462 on Build GRCh38
rs781459943 Chr.8: 31058521-31058521 on Build GRCh38
rs749390862 Chr.8: 31058547-31058547 on Build GRCh38
rs1024022898 Chr.8: 31058448-31058448 on Build GRCh38
rs754627643 Chr.8: 31058520-31058520 on Build GRCh38
rs373503267 Chr.8: 31058484-31058484 on Build GRCh38
rs202148988 Chr.8: 31058463-31058463 on Build GRCh38
rs888468405 Chr.8: 31058475-31058475 on Build GRCh38
rs778355874 Chr.8: 31058526-31058526 on Build GRCh38
rs3087420 Chr.8: 31058489-31058489 on Build GRCh38
rs1006884728 Chr.8: 31058485-31058485 on Build GRCh38
rs550926459 Chr.8: 31058524-31058524 on Build GRCh38
rs761784221 Chr.8: 31058459-31058459 on Build GRCh38
rs757791580 Chr.8: 31058472-31058472 on Build GRCh38
rs34477820 Chr.8: 31058542-31058542 on Build GRCh38
rs538314935 Chr.8: 31058529-31058529 on Build GRCh38

View Full Product Details