Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13922155-13922313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6838968 c.357G>T p.L119F Chr.16: 13922180-13922180 on Build GRCh38
967190 c.354C>A p.F118L Chr.16: 13922177-13922177 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373361309 Chr.16: 13922231-13922236 on Build GRCh38
rs761146709 Chr.16: 13922253-13922254 on Build GRCh38
rs768649885 Chr.16: 13922231-13922231 on Build GRCh38
rs555020322 Chr.16: 13922186-13922186 on Build GRCh38
rs113434368 Chr.16: 13922253-13922253 on Build GRCh38
rs763620308 Chr.16: 13922253-13922255 on Build GRCh38
rs918305286 Chr.16: 13922227-13922227 on Build GRCh38
rs772418764 Chr.16: 13922252-13922253 on Build GRCh38
rs747613537 Chr.16: 13922239-13922239 on Build GRCh38
rs941012957 Chr.16: 13922200-13922200 on Build GRCh38
rs746393376 Chr.16: 13922232-13922233 on Build GRCh38
rs1015821969 Chr.16: 13922265-13922265 on Build GRCh38
rs769539825 Chr.16: 13922252-13922252 on Build GRCh38
rs774988274 Chr.16: 13922228-13922233 on Build GRCh38
rs575078496 Chr.16: 13922278-13922278 on Build GRCh38
rs772713662 Chr.16: 13922258-13922258 on Build GRCh38
rs1038044252 Chr.16: 13922226-13922226 on Build GRCh38
rs763445140 Chr.16: 13922263-13922263 on Build GRCh38
rs746957529 Chr.16: 13922230-13922230 on Build GRCh38
rs776312000 Chr.16: 13922237-13922237 on Build GRCh38
rs34861763 Chr.16: 13922264-13922265 on Build GRCh38
rs144666685 Chr.16: 13922190-13922190 on Build GRCh38
rs777834718 Chr.16: 13922183-13922183 on Build GRCh38
rs780096152 Chr.16: 13922195-13922195 on Build GRCh38

View Full Product Details