The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316364.1 | NP_001303293.1 | 3 - 4 | 533 | 148 |
NM_003908.4 | NP_003899.2 | 3 - 4 | 533 | 148 | |
XM_017028118.1 | XP_016883607.1 | 3 - 4 | 695 | 148 | |
GenBank mRNA | AB451237.1 | - | 2 - 3 | 287 | 148 |
AB451362.1 | - | 2 - 3 | 287 | 148 | |
AK313025.1 | - | 3 - 4 | 404 | 148 | |
BC000461.2 | - | 3 - 4 | 423 | 148 | |
BC000934.2 | - | 3 - 4 | 398 | 148 | |
BC007888.2 | - | 2 - 3 | 214 | 148 | |
BT009795.1 | - | 2 - 3 | 287 | 148 | |
BX418731.2 | - | 3 - 4 | 403 | 148 | |
CR456734.1 | - | 2 - 3 | 287 | 148 | |
M29536.1 | - | 3 - 4 | 403 | 148 |