The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014282.3 | NP_055097.2 | 3 - 4 | 796 | 77 |
XM_005251812.3 | XP_005251869.1 | 3 - 4 | 627 | 77 | |
XM_017014484.1 | XP_016869973.1 | 3 - 4 | 458 | 77 | |
GenBank mRNA | AF241831.1 | - | 3 - 4 | 746 | 77 |
AK025144.1 | - | 1 - 2 | 143 | 77 | |
BQ962055.1 | - | 3 - 4 | 511 | 77 | |
U77327.1 | - | 2 - 3 | 329 | 77 |