The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198558.1 | NP_001185487.1 | 4 - 5 | 1110 | 65 |
NM_014441.2 | NP_055256.1 | 4 - 5 | 1110 | 65 | |
XM_006723146.3 | XP_006723209.1 | 4 - 5 | 1082 | 65 | |
XM_011526730.2 | XP_011525032.1 | 4 - 5 | 1082 | 65 | |
XM_011526732.2 | XP_011525034.1 | 4 - 5 | 1082 | 65 | |
XM_017026595.1 | XP_016882084.1 | 4 - 5 | 1082 | 65 | |
XM_017026596.1 | XP_016882085.1 | 4 - 5 | 1082 | 65 | |
GenBank mRNA | AB026265.1 | - | 4 - 5 | 1061 | 65 |
AF227924.1 | - | 4 - 5 | 1082 | 65 | |
AF247180.1 | - | 4 - 5 | 1026 | 65 | |
AK297043.1 | - | 4 - 5 | 1082 | 65 | |
AK315272.1 | - | 4 - 5 | 1052 | 65 | |
AY358913.1 | - | 4 - 5 | 1057 | 65 | |
BC035365.1 | - | 4 - 5 | 1064 | 65 |