Product Details

Assay Reference Genome
Location

Chr.17:7688979 on build GRCh38
Cytoband
17p13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55135
Gene Symbol
WRAP53
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
Location
Chr.17:7686071-7703502 on build GRCh38
Assay Gene Location
Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
WRAP53 NM_001143990.1 2 486 NP_001137462.1
NM_001143991.1 2 496 NP_001137463.1
NM_001143992.1 2 545 NP_001137464.1
NM_018081.2 1 631 NP_060551.2
XM_011523952.2 1 279 XP_011522254.1
XM_017024812.1 2 1322 XP_016880301.1
AK001247.1 1 503 BAA91579.1
AK056669.1 1 2300
AK092370.1 2 1294
AK225273.1 1 503
AK225411.1 1 503
AK303246.1
AY766322.1 2 496
BC002336.2 2 406 AAH02336.1
DQ431240.1 2 545
DQ431241.1 2 486

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv574322 Chr17:7010333 - 7689462 on Build GRCh38 Loss CLDN7 CHRNB1 ALOX12 MIR324 PLSCR3 SHBG C17orf49 KCTD11 POLR2A SPEM1 ALOX12-AS1 ACADVL C17orf74 SNORA67 NLGN2 CD68 ACAP1 MPDU1 FXR2 SENP3-EIF4A1 TP53 GABARAP DVL2 DLG4 TNFSF12 TNFSF12-TNFSF13 YBX2 TNFSF13 SLC16A13 NEURL4 TMEM256 EIF5A MIR497HG SENP3 ZBTB4 BCL6B TNK1 SLC16A11 LOC107983988 GPS2 MIR195 ATP1B2 ELP5 SNORD10 TMEM102 SOX15 SLC2A4 TMEM256-PLSCR3 SNORA48 ASGR2 SAT2 CTDNEP1 PHF23 WRAP53 CLEC10A TMEM95 RNASEK MIR497 FGF11 RNASEK-C17orf49 LOC100996842 EIF4A1 ASGR1 SLC35G6

View Full Product Details