The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018847.2 | NP_061335.1 | 1 - 1 | 410 | 56 |
GenBank mRNA | AB037775.1 | - | 1 - 1 | 349 | 56 |
AK091715.1 | - | 1 - 1 | 360 | 56 | |
AL713669.1 | - | 1 - 1 | 352 | 56 | |
AY172949.1 | - | 1 - 1 | 369 | 56 | |
BC039133.2 | - | 1 - 1 | 353 | 56 |