Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN4X
Entrez Gene ID
57502
Gene Name
neuroligin 4, X-linked
Gene Aliases
ASPGX2, AUTSX2, HLNX, HNL4X, NLGN4
Chromosome Location
Chr.X: 5890026 - 6228882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020742.3 NP_065793.1 1 - 2 158 66
GenBank mRNA AB033086.1 - 1 - 2 158 66
AF376803.1 - 1 - 2 158 66
AX773938.1 - 1 - 2 158 66

View Full Product Details