The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247987.1 | NP_001234916.1 | 6 - 7 | 1192 | 107 |
NM_020826.2 | NP_065877.1 | 4 - 5 | 974 | 107 | |
GenBank mRNA | AB037848.1 | - | 4 - 5 | 960 | 107 |
AJ303362.1 | - | 2 - 3 | 401 | 107 | |
AK291009.1 | - | 4 - 5 | 895 | 107 | |
AK308841.1 | - | 6 - 7 | 926 | 107 | |
AL512743.1 | - | 4 - 5 | 897 | 107 | |
BC093830.1 | - | 4 - 5 | 894 | 107 | |
BC093832.1 | - | 4 - 5 | 896 | 107 |