The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021129.3 | NP_066952.1 | 6 - 7 | 606 | 130 |
GenBank mRNA | AB026723.1 | - | 6 - 7 | 529 | 130 |
AF092439.1 | - | 6 - 7 | 576 | 130 | |
AF108211.1 | - | 6 - 7 | 487 | 130 | |
AF119665.1 | - | 6 - 7 | 576 | 130 | |
AF154065.1 | - | 6 - 7 | 544 | 130 | |
AF217186.1 | - | 6 - 7 | 499 | 130 | |
AK058193.1 | - | 6 - 7 | 599 | 130 | |
BC001022.2 | - | 6 - 7 | 581 | 130 | |
BC061581.1 | - | 6 - 7 | 589 | 130 | |
BC105034.1 | - | 6 - 7 | 572 | 130 | |
BC105036.1 | - | 6 - 7 | 572 | 130 | |
BC107882.1 | - | 6 - 7 | 540 | 130 | |
BU520836.1 | - | 6 - 7 | 587 | 130 | |
EU794626.1 | - | 6 - 7 | 606 | 130 |