The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021129.3 | NP_066952.1 | 9 - 10 | 900 | 78 |
GenBank mRNA | AB026723.1 | - | 9 - 10 | 823 | 78 |
AF092439.1 | - | 9 - 10 | 870 | 78 | |
AF119665.1 | - | 9 - 10 | 870 | 78 | |
AF154065.1 | - | 9 - 10 | 838 | 78 | |
AF217186.1 | - | 9 - 10 | 793 | 78 | |
AK058193.1 | - | 9 - 10 | 893 | 78 | |
BC001022.2 | - | 9 - 10 | 875 | 78 | |
BC061581.1 | - | 9 - 10 | 883 | 78 | |
BC105034.1 | - | 9 - 10 | 866 | 78 | |
BC105036.1 | - | 9 - 10 | 866 | 78 | |
BC107882.1 | - | 9 - 10 | 834 | 78 | |
EU794626.1 | - | 9 - 10 | 900 | 78 |